Skip to Content
Merck

Skip To

39082

Lead Standard for ICP

TraceCERT®, 10 g/L Pb in nitric acid (nominal concentration)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

product line

TraceCERT®

grade

certified reference material, TraceCERT®

concentration

10 g/L Pb in nitric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Search & download your certificate here.

Application

Lead Standard can be used as a calibration standard in elemental analysis by Inductively coupled plasma Optical Emission spectrometry (ICP-OES) method.

Preparation Note

10 g/L Pb in 5% nitric acid, prepared with high purity Pb(NO3)2 , HNO3 and water

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
413185177775015
grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

technique(s)

ICP: suitable

technique(s)

ICP: suitable

technique(s)

ion chromatography: suitable

technique(s)

ICP: suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

product line

TraceCERT®

product line

TraceCERT®

product line

TraceCERT®

product line

TraceCERT®

application(s)

industrial qc
pharmaceutical

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

environmental
food and beverages

application(s)

industrial qc
pharmaceutical

concentration

10 g/L Pb in nitric acid (nominal concentration)

concentration

1 g/L Pb in nitric acid (nominal concentration)

concentration

1000 mg/L Pb2+ in nitric acid (nominal concentration)

concentration

1 mg/L Pb in nitric acid (nominal concentration)


Still not finding the right product?

Explore all of our products under Lead Standard for ICP


signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1A - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2

target_organs

Blood,Central nervous system,Immune system,Kidney

supp_hazards

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
39082-100ML04061831972979

Questions

Reviews

No rating value

Active Filters