Skip to Content
Merck

Skip To

33799

Isoxadifen-ethyl

PESTANAL®, analytical standard

Synonym(s):

Ethyl 4,5-dihydro-5,5-diphenylisoxazol-3-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H17NO3
CAS Number:
Molecular Weight:
295.33
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
443-870-0
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)C1=NOC(C1)(c2ccccc2)c3ccccc3

InChI

1S/C18H17NO3/c1-2-21-17(20)16-13-18(22-19-16,14-9-5-3-6-10-14)15-11-7-4-8-12-15/h3-12H,2,13H2,1H3

InChI key

MWKVXOJATACCCH-UHFFFAOYSA-N

General description

Isoxadifen-ethyl can play the role as a safener with pesticide fenoxaprop-ethyl, which can be an effective combination in agriculture to protect rice. It can also be used as a herbicide for cereal crops to combat grassy weeds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CRM337993792346302
format

neat

format

-

format

neat

format

neat

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

analytical standard

grade

certified reference material, TraceCERT®

grade

analytical standard

grade

analytical standard

product line

PESTANAL®

product line

TraceCERT®

product line

PESTANAL®

product line

PESTANAL®

application(s)

agriculture
environmental

application(s)

-

application(s)

agriculture
environmental

application(s)

agriculture
environmental

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


Still not finding the right product?

Explore all of our products under Isoxadifen-ethyl


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



4?Hydroxyphenylpyruvate Dioxygenase Inhibitors in Combination with Safeners: Solutions for Modern and Sustainable Agriculture
Ahrens H, et al.
Angewandte Chemie (International Edition in English), 52, 9388-9398 (2013)
Pesticides?Advances in Research and Application (2013)
4-Hydroxyphenylpyruvate Dioxygenase Inhibitors in Combination with Safeners: Solutions for Modern and Sustainable Agriculture
Ahrens H, et al.
Angewandte Chemie (International Edition in English), 52(36), 9388-9398 (2013)



Global Trade Item Number

SKUGTIN
33799-100MG04061826744062

Questions

Reviews

No rating value

Active Filters