Skip to Content
Merck

Skip To

31301

Di-n-octyl phthalate

analytical standard

Synonym(s):

DNOP, Phthalic acid di-n-octyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
204-214-7
Beilstein/REAXYS Number:
1915994
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

density

0.980 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC

InChI

1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3

InChI key

MQIUGAXCHLFZKX-UHFFFAOYSA-N

General description

Di-n-octyl phthalate, belonging to the class of plasticizers, is found to be omnipresent in the environment. It is reported to be used in the manufacturing of flooring tiles, bottle cap liners, indirect food additives and toys.
Di-n-octyl phthalate (DNOP) is a mixed component of C6–C10 linear-chain phthalates which has the ability to impart flexibility in plastics and rubber materials. It is also used as plasticizer in PVC used in flooring and carpet tiles.

Application

Di-n-octyl phthalate may be used as an analytical reference standard for the quantification of the analyte in:
  • Soil samples using micellar electro-kinetic chromatography (MEKC), further confirmed by gas chromatography with flame ionization detection (GC-FID).
  • Bottled water using gas chromatography coupled to mass spectrometry (GC-MS).
  • Plastic toys using gas chromatography with flame ionization detection (GC-FID).

DNOP has been used as standard during solid-phase microextraction followed by GC-MS analysis of phthalate esters in water sample examined after sonochemical degradation.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
881738015336736
grade

analytical standard

grade

certified reference material, TraceCERT®

grade

-

grade

analytical standard

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

environmental

application(s)

-

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

format

neat

format

-

format

neat

density

0.980 g/mL at 20 °C (lit.)

density

0.980 g/mL at 20 °C (lit.)

density

0.980 g/mL at 20 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

100


Still not finding the right product?

Explore all of our products under Di-n-octyl phthalate


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

228.2 °F - closed cup

flash_point_c

109.0 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters