Skip to Content
Merck

Skip To

BCIPP-RO

Roche

BCIP

99% (HPLC), powder, solubility: in DMF, soluble

Synonym(s):

5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt, BCIP® p-toluidine salt, X-phosphate p-toluidine salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H6BrClNO4P · C7H9N
CAS Number:
Molecular Weight:
433.62
UNSPSC Code:
12352204
PubChem Substance ID:
MDL number:
NACRES:
NA.54
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

5-bromo-4-chloro-3-indolyl-phosphate, 4-toluidine salt

assay

99% (HPLC)

form

powder

mol wt

Mr 326.44 (BCIP), Mr 433.6 (BCIP toluidine salt)

packaging

pkg of 1 g (11585002001), pkg of 250 mg (10760994001)

manufacturer/tradename

Roche

solubility

DMF: soluble

storage temp.

2-8°C

SMILES string

Cc1ccc(N)cc1.OP(O)(=O)Oc2c[nH]c3ccc(Br)c(Cl)c23

InChI

1S/C8H6BrClNO4P.C7H9N/c9-4-1-2-5-7(8(4)10)6(3-11-5)15-16(12,13)14;1-6-2-4-7(8)5-3-6/h1-3,11H,(H2,12,13,14);2-5H,8H2,1H3

InChI key

QEIFSLUFHRCVQL-UHFFFAOYSA-N

General description

BCIP serves as substrate for alkaline phosphatase. The reaction product has a blue color and is insoluble in water. The blue color can be visually detected.
Histochemical substrate for alkaline phosphatase.

Application

For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.
BCIP is provided in two salt forms: the disodium salt which is soluble in water and the p-toluidine form which is soluble in dimethylformamide. These salt forms may be used to prepare a stock solution that in combination with NBT and a reaction buffer, form a substrate solution for alkaline phosphatase. This substrate system, when incubated with alkaline phosphatase, produces an insoluble NBT diformazan product that is easily observable with its purple color. For added convenience, a mixture of BCIP and NBT is provided in an easily dissolvable tablet form or as a ready-to-use liquid.
For the colorimetric detection of alkaline phosphatase-labeled molecules.

Analysis Note

Performance test: incubation with AP
Absorption: Absorption maximum in the visible light is not determined.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1168145100111383221001HPA013562
assay

99% (HPLC)

assay

-

assay

>95% (HPLC)

assay

-

solubility

DMF: soluble

solubility

-

solubility

-

solubility

-

form

powder

form

solution

form

solution

form

buffered aqueous glycerol solution

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

mol wt

Mr 326.44 (BCIP), Mr 433.6 (BCIP toluidine salt)

mol wt

817.7

mol wt

-

mol wt

-

description

5-bromo-4-chloro-3-indolyl-phosphate, 4-toluidine salt

description

-

description

4-toluidine salt

description

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

136.4 °F

flash_point_c

58 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
1076099400104061838675408
1158500200104061838251077