Skip to Content
Merck

Skip To

487666

p-Nitrophenyl Phosphate Tablets

Excellent substrate for alkaline and acid phosphatase-based ELISA assays.

Synonym(s):

p-Nitrophenyl Phosphate Tablets, pNPP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.77
UNSPSC Code:
41116158
Form:
(24 Tablets)
Quality level:
Storage condition:
OK to freeze
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

(24 Tablets)

packaging

pkg of 24 tablets

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

General description

Each tablet contains 5 mg pNPP.
Excellent substrate for alkaline and acid phosphatase-based ELISA assays. Produces a yellow soluble end product that can be read at 405 nm. Both kinetic and endpoint determinations can be performed.
Excellent substrate for alkaline and phosphatase-based ELISA assays. Produces a yellow soluble end product that can be read at 405 nm. Both kinetic and endpoint determinations can be performed. Each tablet contains 5 mg pNPP.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4876554876487870
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze, protect from light

storage condition

OK to freeze

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

packaging

pkg of 24 tablets

packaging

-

packaging

-

packaging

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
487666-1EA04055977200300

Questions

  1. Provide the preparation note for this product. (Usage Instructions)

    1 answer
    1. These 5 mg pNPP tablets can be dissolved in water or aqueous buffer solutions (e.g., 50 mM Tris buffer, pH 7.0). For a typical ELISA assay, a working concentration range of 1 to 10 mg/ml is generally used, with the stock concentration being 5 to 10 times its working concentration. Preparing a 0.5 ml solution with one tablet results in a concentration of 10 mg/ml.

      Helpful?

Reviews

No rating value

Active Filters