Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
agency
NF, Ph. Eur.
product line
EMPROVE® EXPERT
form
liquid
density
1.053 g/cm3 at 20 °C
application(s)
pharmaceutical
storage temp.
2-30°C
SMILES string
[P](=O)([O-])([O-])[O-].[H+].[H+].[H+]
InChI
1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)
InChI key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
General description
Application
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form - | form liquid |
| application(s) pharmaceutical | application(s) liquid formulation | application(s) pharmaceutical small molecule | application(s) - |
| agency NF, Ph. Eur. | agency BP, ChP, JPE, NF, Ph. Eur. | agency traceable to USP 1535802 | agency reag. ISO, reag. Ph. Eur. |
| storage temp. 2-30°C | storage temp. 2-30°C | storage temp. 2-30°C | storage temp. no temp limit |
| Quality Level 500 | Quality Level 500 | Quality Level 300 | Quality Level 300 |
| density 1.053 g/cm3 at 20 °C | density 1.685 g/mL at 25 °C (lit.) | density 1.685 g/mL at 25 °C (lit.) | density 1.685 g/mL at 25 °C (lit.) |
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
flash_point_f
Not applicable
flash_point_c
Not applicable
hcodes
Storage Class
12 - Non Combustible Liquids
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


