Skip to Content
Merck

Skip To

1.59010

Ethyl alcohol, pure

greener alternative

92.6-95.2% (m/m), reagent grade,for extraction,for analytical purposes, 95.1-96.9 % (v/v), reag. Ph. Eur., EMSURE®

Synonym(s):

Ethanol, Ethyl alcohol, EtOH, Ethyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
UNSPSC Code:
12191502
EC Index Number:
200-578-6
NACRES:
NB.21
Beilstein/REAXYS Number:
1718733
MDL number:
Assay:
92.6-95.2% (m/m)
Bp:
78 °C (lit.)
Vapor pressure:
44.6 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ethanol 96%, EMSURE® Reag. Ph Eur

agency

reag. Ph. Eur.

Quality Segment

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

description

for analytical purposes, for extraction

product line

EMSURE®

assay

92.6-95.2% (m/m)

form

liquid

autoignition temp.

683 °F

potency

6200 mg/kg LD50, oral (Rat)

expl. lim.

19 %, 60 °F

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

95.1-96.9 % (v/v)

dilution

(for analytical testing)

evapn. residue

≤25 mg/L

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L in H2O)

bp

78 °C (lit.)

mp

−114 °C (lit.)

transition temp

flash point 17 °C

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

greener alternative category

storage temp.

2-30°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biorenewable and alternative to petrochemical solvents. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here  for more information.

Application

Ethyl alcohol, pure can be used as a solvent in the extraction of:
  • Bio-pigments from the green microalgae Chlorella pyrenoidosa.
  • Cannabinoids and terpenoids in Cannabis sativa.
  • Phenolic compounds from Sacha Inchi Shell.

Other Notes

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration
Discover EMSURE - to be more than sure!
Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.009831.070178.18760
assay

92.6-95.2% (m/m)

assay

≥99.9% (GC)

assay

≥99.5% (GC)

assay

≥99.5% (GC)

Quality Level

300

Quality Level

300

Quality Level

200

Quality Level

200

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

application(s)

environmental
food and beverages
industrial qc
pharmaceutical

form

liquid

form

liquid

form

liquid

form

liquid

refractive index

n20/D 1.3600 (lit.)

refractive index

n20/D 1.36 (lit.)

refractive index

n20/D 1.3600 (lit.)

refractive index

n20/D 1.3600 (lit.)

bp

78 °C (lit.)

bp

78 °C (lit.), 78.29 °C/1013 hPa

bp

78 °C (lit.)

bp

78 °C (lit.)


Still not finding the right product?

Explore all of our products under Ethanol 96%


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

62.6 °F

flash_point_c

17 °C

hcodes



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. This can be used for Perfumery?

    1 answer
    1. This product is intended for research use only and is not suitable for use in Fragrance or Perfumery applications. This material is not suitable for in vitro diagnostics or therapeutic use in humans or animals.

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. Can it be used to make dyes, is it denatured?

    1 answer
    1. Yes, this product can be used as a solvent to make dyes. It is not denatured.

      Helpful?

Reviews

No rating value

Active Filters