Skip to Content
Merck

Skip To

1.37017

Sodium chloride

EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP

Synonym(s):

Halite, NaCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
EC Index Number:
231-598-3
UNSPSC Code:
12352302
Beilstein/REAXYS Number:
3534976
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

agency

BP, ChP, JP, Ph. Eur., USP

vapor pressure

1.3 hPa ( 865 °C)

product line

EMPROVE® EXPERT

form

solid

feature

RNase and DNase free

pH

4.5-7.0 (20 °C, 100 g/L in H2O)

bp

1461 °C/1013 hPa

mp

801 °C (lit.)

density

2.17 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

2-30°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium chloride is a mineral salt with broad range of applications in pharmaceutical formulations. It is typically used as an osmotic agent in liquid dosage forms e.g. for preparation of isotonic formulations, as bulking or coating agent for solid dosage forms, as well as for semi-solid dosage forms like creams.
Due to its low microbial and endotoxin limits, Sodium chloride Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.162241.041631613804
application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical
solid formulation

application(s)

pharmaceutical (small molecule)

agency

BP, JP, USP, ChP, Ph. Eur.

agency

BP, JP, Ph. Eur., USP

agency

BP, ChP, JP, Ph. Eur., USP

agency

-

form

solid

form

solid

form

granular

form

-

density

2.17 g/cm3 at 20 °C

density

2.17 g/cm3 at 20 °C

density

-

density

-

product line

EMPROVE® EXPERT

product line

EMPROVE® EXPERT

product line

EMPROVE® EXPERT

product line

-

vapor pressure

1.3 hPa ( 865 °C)

vapor pressure

1.3 hPa ( 865 °C)

vapor pressure

-

vapor pressure

1 mmHg ( 865 °C)


Still not finding the right product?

Explore all of our products under Sodium chloride


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
137017000504054839392511
137017901204027269077631
137017000004027269071639
137017701204065274141053
137017000104027269365295
137017950004027269354060
137017500004027269071646
137017905104065272079037
137017100004027269292805
137017902904027269071653
137017999904027269071660
137017602504027269679996
137017910004054839392528

Questions

Reviews

No rating value

Active Filters