Skip to Content
Merck

Skip To

1.37010

di-Potassium hydrogen phosphate

anhydrous, EMPROVE® EXPERT, Ph. Eur., BP, USP

Synonym(s):

Potassium phosphate dibasic, Dipotassium hydrogen phosphate, Potassium phosphate dibasic, Dipotassium Phosphate, Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
UNSPSC Code:
12352302
EC Index Number:
231-834-5
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, Ph. Eur., USP

product line

EMPROVE® EXPERT

form

solid

feature

RNase and DNase free

pH

9 (20 °C, 10 g/L in H2O)

solubility

1600 g/L

density

2.44 g/cm3 at 20 °C

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

15-25°C

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

di-Potassium hydrogen phosphate anhydrous is typically used as a phosphate buffer system component for biomolecule downstream chromatography steps or final liquid formulation.
Due to its low microbial and endotoxin limits, di-Potassium hydrogen phosphate anhydrous Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.051011551128PHR2900
application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

agency

BP, USP, Ph. Eur.

agency

BP, Ph. Eur.

agency

-

agency

traceable to USP 1551128

form

solid

form

solid

form

-

form

-

solubility

1600 g/L

solubility

1600 g/L

solubility

-

solubility

-

product line

EMPROVE® EXPERT

product line

EMPROVE® ESSENTIAL

product line

-

product line

-

density

2.44 g/cm3 at 20 °C

density

2.44 g/cm3 at 20 °C

density

-

density

-


Still not finding the right product?

Explore all of our products under di-Potassium hydrogen phosphate


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
137010628604065271284159
137010902904027269059972
137010679204065271284166
137010910004054839029370
137010000104027269279653
137010070104065271284180
137010100004027269303402
137010070204065271284197
137010070304065271284203
137010680304065271334526
137010500004027269059965
137010901204027269077495

Questions

Reviews

No rating value

Active Filters