Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Segment
agency
BP, ChP, JP, Ph. Eur., USP
vapor pressure
<1 Pa ( 25 °C)
description
cryst. EMPROVE® EXPERT Ph Eur,BP,ChP,JP,USP
product line
EMPROVE® EXPERT
form
solid, crystalline
feature
RNase and DNase free
impurities
endotoxins, low
pH
1.85 (25 °C, 50 g/L in H2O)
pKa
(1) 3.13, (2) 4.76, (3) 6.4
mp
135-152 °C
transition temp
flash point 173.9 °C
solubility
880 g/L
density
1.54 g/cm3 at 20 °C
application(s)
liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical
storage temp.
15-25°C
SMILES string
OC(CC(O)(C(O)=O)CC(O)=O)=O.O
InChI
1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2
InChI key
YASYEJJMZJALEJ-UHFFFAOYSA-N
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Due to its low microbial and endotoxin limits, Citric acid anhydrous Emprove® Expert is especially suitable for high-risk applications.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| application(s) liquid formulation | application(s) liquid formulation | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical |
| agency BP, JP, USP, ChP, Ph. Eur. | agency BP, ChP, Ph. Eur., USP | agency EP Reference Standard | agency traceable to Ph. Eur. C2219000, traceable to USP 1134368 |
| form crystalline, solid | form coarse powder, crystalline | form - | form - |
| product line EMPROVE® EXPERT | product line EMPROVE® ESSENTIAL | product line - | product line - |
| solubility 880 g/L | solubility 880 g/L | solubility - | solubility - |
| density 1.54 g/cm3 at 20 °C | density 1.54 g/cm3 at 20 °C | density - | density - |
Still not finding the right product?
Explore all of our products under Citric acid monohydrate
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
345.0 °F - closed cup
flash_point_c
173.9 °C - closed cup
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1370039012 | 04027269077426 |
| 1370030001 | 04027269365233 |
| 1370039029 | 04027269061968 |
| 1370035000 | 04027269061432 |
| 1370030503 | 04027269171865 |
| 1370031000 | 04027269364946 |


