Skip to Content
Merck

Skip To

1.04366

n-Heptane

greener alternative

for spectroscopy Uvasol®

Synonym(s):

Heptane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
UNSPSC Code:
12191502
NACRES:
NA.03
EC Index Number:
205-563-8
MDL number:
Beilstein/REAXYS Number:
1730763
eCl@ss:
39010112
Assay:
≥99.3% (GC)
Technique(s):
UV/Vis spectroscopy: suitable
Bp:
98 °C (lit.)
Vapor pressure:
40 mmHg ( 20 °C), 83 mmHg ( 37.7 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3.5 (vs air)

Quality Level

vapor pressure

40 mmHg ( 20 °C), 83 mmHg ( 37.7 °C)

assay

≥99.3% (GC)

form

liquid

autoignition temp.

433 °F

potency

>5000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0001 meq/g Acidity, ≤0.0002 meq/g Alkalinity, ≤0.005% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

200 nm, ≥20%, 210 nm, ≥55%, 220 nm, ≥80%, 228 nm, ≥90%, 245 nm, ≥98%

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

transition temp

flash point -4 °C

solubility

0.05 g/L

density

0.684 g/mL at 25 °C (lit.)

UV absorption

λ: 200 nm Amax: ≤0.70, λ: 210 nm Amax: ≤0.26, λ: 220 nm Amax: ≤0.10, λ: 228 nm Amax: ≤0.05, λ: 245 nm Amax: ≤0.01

greener alternative category

storage temp.

2-30°C

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Application


  • Cloning, Expression, Characterization and Immobilization of a Recombinant Carboxylesterase from the Halophilic Archaeon, Halobacterium salinarum NCR-1.: This study involves the characterization and immobilization of a recombinant carboxylesterase using n-Heptane as a solvent in the enzyme activity assays, highlighting its potential applications in biocatalysis and industrial processes (Ortega-de la Rosa et al., 2024).

  • Biomonitoring of volatile organic compounds and organophosphorus flame retardants in commercial aircrews after "fume and smell events".: This research employs n-Heptane in analytical methods for detecting volatile organic compounds, underlining its importance in environmental health studies and air quality monitoring (Weiss et al., 2024).

  • Exploring Deactivation Reasons of Biomass-Based Phosphorus-Doped Carbon as a Metal-Free Catalyst in the Catalytic Dehydroaromatization of n-Heptane.: This article examines the deactivation of catalysts in the dehydroaromatization process of n-Heptane, providing insights into catalyst design and optimization for chemical synthesis (Yu et al., 2024).

  • Reversible solvent interactions with UiO-67 metal-organic frameworks.: The study explores the interactions of n-Heptane with metal-organic frameworks, demonstrating its application in materials science and solvent recovery processes (Goodenough et al., 2024).

  • Capillary electrokinetic chromatography for chiral separation of potential SARS-CoV-2 3CL protease inhibitors.: This research uses n-Heptane in capillary electrokinetic chromatography for the chiral separation of pharmaceutical compounds, emphasizing its role in drug development and analytical chemistry (Brier et al., 2024).

Analysis Note

Purity (GC): ≥ 99.3 %
Evaporation residue: ≤ 0.0002 %
Water: ≤ 0.005 %
Color: ≤ 10 Hazen
Acidity: ≤ 0.0001 meq/g
Alkalinity: ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 1.0 ppb
Transmission (at 200 nm): ≥ 20 %
Transmission (at 210 nm): ≥ 55 %
Transmission (at 220 nm): ≥ 80 %
Transmission (at 228 nm): ≥ 90 %
Transmission (from 245 nm): ≥ 98 %
Absorbance (at 200 nm): ≤ 0.70
Absorbance (at 210 nm): ≤ 0.26
Absorbance (at 220 nm): ≤ 0.10
Absorbance (at 228 nm): ≤ 0.05
Absorbance (from 245 nm): ≤ 0.01

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent compared to hexane and thus align with "Safer Solvents and Auxiliaries". Click here for more information.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.043601.043901.04379
assay

≥99.3% (GC)

assay

≥99.5% (GC)

assay

≥99.3% (GC)

assay

≥99% (GC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

300

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

gas chromatography (GC): suitable

technique(s)

HPLC: suitable

technique(s)

-

solubility

0.05 g/L

solubility

-

solubility

0.05 g/L

solubility

-

form

liquid

form

liquid

form

liquid

form

liquid

impurities

≤0.0001 meq/g Acidity, ≤0.005% Water, ≤0.0002 meq/g Alkalinity

impurities

-

impurities

≤0.0002 meq/g Acidity, ≤0.0002 meq/g Alkalinity, ≤0.005% Water

impurities

≤0.0005% Free acid (as CH3COOH), ≤0.005% Sulfur compounds (as S), ≤0.01% Water, ≤0.1% Aromatic hydrocarbons (as C6H6)


Still not finding the right product?

Explore all of our products under n-Heptane


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
104366050004022536050461
104366250004022536050485

Questions

Reviews

No rating value

Active Filters