Skip to Content
Merck

Skip To

1.04352

L-Histidine

EMPROVE® EXPERT, Ph. Eur., ChP, JP, USP

Synonym(s):

His, (S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
EC Index Number:
200-745-3
UNSPSC Code:
12352209
Beilstein/REAXYS Number:
84088
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

agency

ChP, JP, Ph. Eur., USP

product line

EMPROVE® EXPERT

form

solid

technique(s)

API processing | cocrystal formation: suitable

pH

7.7 (20 °C, 10 g/L in H2O)

mp

282 °C (dec.) (lit.)

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

2-30°C

SMILES string

N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1

InChI key

HNDVDQJCIGZPNO-YFKPBYRVSA-N

General description

Chemicals play an important role in the stabilization of a biologic drug during its manufacturing and formulation process – for instance, by preventing aggregation. We offer a wide range of high-quality stabilizers, buffers and salts to successfully purify and formulate your biomolecules. Specifically developed for high-risk applications, they are low in bioburden and endotoxins.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Histidine is used in biopharma manufacturing as a buffer in formulation. It is often used together with histidine monohydrochloride to prepare histidine buffer with the desired pH value.
Due to its low microbial and endotoxin limits, L-Histidine Emprove® Expert is especially suitable for high-risk applications.

Biochem/physiol Actions

Precursor of histamine by action of histidine decarboxylase.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR110873767H3911
application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

pharmaceutical (small molecule)

agency

ChP, Ph. Eur., JP, USP

agency

traceable to Ph. Eur. H0750000, traceable to USP 1308505

agency

-

agency

-

technique(s)

API processing | cocrystal formation: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

cell culture | mammalian: suitable

form

solid

form

-

form

-

form

crystalline powder

product line

EMPROVE® EXPERT

product line

-

product line

TraceCERT®

product line

-

pH

7.7 (20 °C, 10 g/L in H2O)

pH

-

pH

-

pH

-


Still not finding the right product?

Explore all of our products under L-Histidine


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 1



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
104315010004065267115689
104352100004022536906829
104352050004027269365042
104352000104027269365486
104352901004022536906836

Questions

Reviews

No rating value

Active Filters