Skip to Content
Merck

Skip To

1.02406

Sodium chloride

volumetric standard, secondary reference material for argentometry, traceable to NIST SRM Certipur® Reag. Ph Eur,Reag. USP

Synonym(s):

Halite, NaCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
UNSPSC Code:
41116107
NACRES:
NA.24
EC Index Number:
231-598-3
MDL number:
Beilstein/REAXYS Number:
3534976
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

agency

ISO 17025, reag. Ph. Eur., reag. USP

vapor pressure

1.3 hPa ( 865 °C)

product line

Certipur®

form

solid

potency

3000 mg/kg LD50, oral (Rat), >10000 mg/kg LD50, skin (Rabbit)

technique(s)

titration: suitable

pH

4.5-7.0 (20 °C, 100 g/L in H2O)

bp

1461 °C/1013 hPa

mp

801 °C (lit.)

solubility

358 g/L

bulk density

1140 kg/m3

storage temp.

15-25°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

General description

Sodium chloride (NaCl) is a volumetric standard manufactured under rigorous conditions and analyzed with the highest possible precision. The mass fraction of the standard is determined by potentiometric titration, and it is qualified in a DIN EN ISO/IEC 17025 accredited laboratory.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It finds its use as a certified reference material (CRM) for the standardization of volumetric solutions in argentometric titrations, in accordance with USP and Ph. Eur.

Features and Benefits

  • Available as a solid in a contamination-proof glass bottle to ensure its stability for the entire shelf life until opened.
  • Certified reference material according to ISO 17034
  • High-purity material traceable to SRMs from NIST
  • Analyzed in a DIN EN ISO/IEC 17025 accredited laboratory
  • In accordance with the chapter reagents of USP and Ph. Eur
  • High-quality offering accurate titer determinations

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR132194052671380
technique(s)

titration: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

product line

Certipur®

product line

-

product line

-

product line

-

vapor pressure

1.3 hPa ( 865 °C)

vapor pressure

-

vapor pressure

-

vapor pressure

1.33 hPa ( 865 °C)

Quality Level

300

Quality Level

300

Quality Level

100

Quality Level

200

form

solid

form

-

form

crystalline powder

form

powder or crystals

agency

ISO 17025, reag. USP, reag. Ph. Eur.

agency

traceable to USP 1613804

agency

-

agency

-


Still not finding the right product?

Explore all of our products under Sodium chloride


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
102406008004022536555829
102406008304054839654374

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters