Skip to Content
Merck

Skip To

1.01262

Water

for inorganic trace analysis, suitable for UV/Vis spectroscopy, Ultrapur

Synonym(s):

DI water, Deionized water

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
UNSPSC Code:
12352303
NACRES:
NB.21
MDL number:
Beilstein/REAXYS Number:
2050024
Technique(s):
UV/Vis spectroscopy: suitable
Bp:
100 °C (lit.)
Vapor pressure:
23 hPa ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Water, Ultrapur

vapor pressure

23 hPa ( 20 °C)

Quality Level

description

for inorganic trace analysis

form

liquid

technique(s)

UV/Vis spectroscopy: suitable

refractive index

n20/D 1.34 (lit.)

pH

7 (20 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

storage temp.

2-30°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
9601-OP9801-OP4.86505
technique(s)

UV/Vis spectroscopy: suitable

technique(s)

-

technique(s)

-

technique(s)

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

300

form

liquid

form

liquid

form

liquid

form

liquid

density

1.000 g/mL at 3.98 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

bp

100 °C (lit.)

bp

100 °C (lit.)

bp

100 °C (lit.)

bp

100 °C (lit.)

pH

7 (20 °C)

pH

-

pH

-

pH

-


Still not finding the right product?

Explore all of our products under Water


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
101262100004022536509426
101262050004054839514517

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters