Skip to Content
Merck

Skip To

1.00804

L(+)-Tartaric acid

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

L(+)-Tartaric acid, 2,3-Dihydroxybutanedioic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HOOCCH(OH)CH(OH)COOH
CAS Number:
Molecular Weight:
150.09
UNSPSC Code:
12352106
EC Index Number:
201-766-0
NACRES:
NA.21
MDL number:
Assay:
≥99.5-101.0% dry basis (alkalimetric)
Form:
solid
Grade:
ACS reagent
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. ISO, reag. Ph. Eur., suitable for EPA 200.7, suitable for EPA 200.8

vapor pressure

<5 Pa ( 20 °C)

product line

EMSURE®

assay

≥99.5-101.0% dry basis (alkalimetric)

form

solid

autoignition temp.

425 °C

impurities

≤10 ppm Phosphate (PO4), ≤100 ppm Sulfated ash, ≤20 ppm Total sulfur (as SO4), ≤350 ppm Oxalic acid, ≤5 ppm Chloride (Cl), ≤50 ppm Insoluble matter, ≤50 ppm Sulfate (SO4)

loss

≤ 0.2% loss on drying, 105°C

pH

1.6 (25 °C, 100 g/L in H2O)

mp

168-170 °C

transition temp

flash point 210 °C

solubility

water: 1390 g/L at 20 °C

density

1.76 g/cm3 at 20 °C

bulk density

1000 kg/m3

cation traces

Ca: ≤20 ppm, Cu: ≤5 ppm, Fe: ≤5 ppm, Pb: ≤5 ppm, heavy metals (as Pb): ≤5 ppm

storage temp.

2-30°C

SMILES string

[O-]C(=O)C(O)C(O)C(=O)[O-].[H+].[H+]

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)

InChI key

FEWJPZIEWOKRBE-UHFFFAOYSA-N

Application


  • Binary Mixtures of Meloxicam and L-Tartaric Acid for Oral Bioavailability Modulation of Pharmaceutical Dosage Forms.: This study explores the use of L(+)-tartaric acid in binary mixtures to enhance the oral bioavailability of meloxicam, suggesting potential applications in developing more effective pharmaceutical formulations (Macasoi et al., 2024).

  • Hyper-Raman spectroscopy of biomolecules.: This research employs Hyper-Raman spectroscopy, an analytical technique enhanced by L(+)-tartaric acid, to provide insights into the structural dynamics of biomolecules, showcasing its utility in complex biological systems (Marble et al., 2024).

  • Novel multi-component crystals of berberine with improved pharmaceutical properties.: This study highlights the use of L(+)-tartaric acid in the development of novel multi-component crystals, potentially leading to pharmaceuticals with enhanced efficacy and stability (Zhang et al., 2023).

  • Deep Eutectic Solvent (DES)-Mediated One-Pot Multicomponent Green Approach for Naphthalimide-Centered Acridine-1,8-dione Derivatives and Their Photophysical Properties.: Research demonstrates the synthesis of complex organic compounds using L(+)-tartaric acid as a component of a deep eutectic solvent, highlighting its role in green chemistry (Rather et al., 2022).

Analysis Note

Assay (alkalimetric): ≥ 99.5 %
Assay (alkalimetric, calculated on dried substance): 99.5 - 101.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 50 ppm
Spec. rotation [α²0/D (20 %; water calculated on dried substance): 12.0 - 12.8 °
Chloride (Cl): ≤ 5 ppm
Phosphate (PO₄): ≤ 10 ppm
Sulfate (SO₄): ≤ 50 ppm
Heavy metals (as Pb): ≤ 5 ppm
Total sulfur (as SO₄): ≤ 20 ppm
Ca (Calcium): ≤ 20 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Oxalate (C₂O₄): passes test
Oxalic acid: ≤ 350 ppm
Sulfated ash: ≤ 100 ppm
Loss on Drying (105°C): ≤ 0.2 %
Corresponds to ACS,ISO,Reag. Ph Eur

Other Notes

Replaces: TX0010-1; TX0010

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.008021.0080333801
grade

ACS reagent

grade

-

grade

-

grade

-

assay

≥99.5-101.0% dry basis (alkalimetric)

assay

99.7-100.5% dry basis (acidimetric)

assay

99.7-100.5% dry basis (acidimetric)

assay

99.5-101.0% (calc. to the dried substance)

agency

reag. ISO, suitable for EPA 200.7, reag. Ph. Eur., suitable for EPA 200.8

agency

BP, ChP, JP, NF, Ph. Eur.

agency

BP, ChP, JP, NF, Ph. Eur.

agency

Ph. Eur., USP/NF, reag. ISO

Quality Level

300

Quality Level

500

Quality Level

500

Quality Level

200

form

solid

form

coarse powder, crystalline

form

powder

form

powder or crystals

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-


Still not finding the right product?

Explore all of our products under L(+)-Tartaric acid


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters