Density is not part of the tests to release the product and is therefore not measured.
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
non-animal source
Quality Level
agency
EP, JP, USP
sterility
irradiated
form
granular
feature
granulated
packaging
pkg of 5 kg, pkg of 500 g
manufacturer/tradename
GranuCult® prime
technique(s)
media fill: suitable, microbiological culture: suitable
pH
7.1-7.5 (25 °C, 30 g/L in H2O)
solubility
30 g/L
application(s)
aseptic process simulation, pharmaceutical
compatibility
stand alone
storage temp.
15-25°C
suitability
anaerobic bacteria
General description
Application
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| form granular | form granular | form liquid | form liquid (ready-to-use) |
| technique(s) media fill: suitable, microbiological culture: suitable | technique(s) microbiological culture: suitable | technique(s) - | technique(s) - |
| suitability anaerobic bacteria | suitability Cronobacter spp. (spp.), Escherichia coli (pathogenic), Salmonella spp. (spp.), anaerobic bacteria, nonselective for | suitability nonselective for | suitability nonselective for |
| application(s) aseptic process simulation, pharmaceutical | application(s) microbiology, pharmaceutical | application(s) cosmetics | application(s) aseptic process simulation |
| manufacturer/tradename GranuCult® prime | manufacturer/tradename GranuCult® plus | manufacturer/tradename - | manufacturer/tradename - |
| agency EP, USP, JP | agency EP, USP | agency EP 2.6.1, JP 4.06, USP 71 | agency EP, USP |
Still not finding the right product?
Explore all of our products under Tryptic Soy Broth - Dehydrated Culture Media
signalword
Warning
hcodes
Hazard Classifications
Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P66807-100G | 04061833077405 |
| P66807-5G | 04061834393931 |
| P66807-1KG | 04061826686355 |
| 1005509010 | 04065273835212 |
| 1005500500 | 04022536895628 |
| 1005505000 | 04022536895642 |
-
Which is the Density value of the 30 g/L solution of GranuCult® prime Tryptic Soy Broth (TSB) non-animal origin, irradiated (Ord. nr. 1.00550.0500/ 1.00550.5000)?
1 answer-
Helpful?
-




