Skip to Content
Merck

Skip To

1.00165

Boric acid

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Orthoboric acid, Trihydroxidoboron

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
NACRES:
NA.21
EC Index Number:
233-139-2
UNSPSC Code:
12352301
MDL number:
Assay:
99.5-100.5% (alkalimetric)
Concentration:
99.5-100.5%
Form:
solid
Grade:
ACS reagent
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. ISO, reag. Ph. Eur., suitable for SM 4500 - NH3, suitable for SM 5210

vapor pressure

2.6 mmHg ( 20 °C)

product line

EMSURE®

assay

99.5-100.5% (alkalimetric)

form

solid

potency

2660 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

concentration

99.5-100.5%

impurities

≤0.005% In methanol insoluble matter, ≤0.05% Nonvolatile with methanol

pH

3.8-4.8 (20 °C, 33 g/L in H2O)

mp

160 °C (dec.) (lit.)

solubility

49.2 g/L

density

1.489 g/cm3 at 23 °C

bulk density

400‑600 kg/m3

anion traces

chloride (Cl-): ≤0.0003%, phosphate (PO43-): ≤0.0005%, sulfate (SO42-): ≤0.0005%

cation traces

Ca: ≤0.002%, Fe: ≤0.0001%, Pb: ≤0.001%, heavy metals: ≤0.0005% ((ACS))

storage temp.

2-30°C

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

General description

Boric acid (EMSURE® grade product) is used for regulated analyses and highly demanding lab applications. It also offers the highest quality and unmatched scope of specifications to give complete control of test conditions and eliminate uncertainties. EMSURE® grade products comply with all international regulations and can be used in various applications.

Application

Boric acid can be used:
  • For the determination of the protein content of yogurt samples.
  • In the proximate analysis of seaweed Caulerpa sp. samples.

Features and Benefits

  • Most extensive specifications worldwide
  • Tested for up to 70 parameters
  • Extraordinary purity
  • Very low limiting values
  • Greater accuracy and control of analyses
  • Optimized for highly critical and demanding analyses
  • Ideal for method development
  • No interference or contamination due to unknown impurities

Analysis Note

Assay (alkalimetric): 99.5 - 100.5 %


Identity: passes test


Appearance of solution: passes test


pH-value (3.3 %, water): 3.8 - 4.8


Chloride (Cl): ≤ 0.0003 %


Phosphate (PO₄): ≤ 0.0005 %


Sulfate (SO₄): ≤ 0.0005 %


Solubility in ethanol: passes test


In methanol insoluble matter: ≤ 0.005 %


heavy metals (ACS) ≤ 0.0005%


Ca (Calcium): ≤ 0.002 %


Fe (Iron): ≤ 0.0001 %


Pb (Lead): ≤ 0.001 %


Organic impurities: passes test


Nonvolatile with methanol: ≤ 0.05 %


Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
107634131146NIST973
grade

ACS reagent

grade

pharmaceutical primary standard

grade

ACS reagent, puriss. p.a.

grade

certified reference material

concentration

99.5-100.5%

concentration

-

concentration

-

concentration

-

assay

99.5-100.5% (alkalimetric)

assay

-

assay

≥99.8%

assay

-

agency

reag. ISO, suitable for SM 4500 - NH3, reag. Ph. Eur., suitable for SM 5210

agency

-

agency

USP/NF, reag. ISO, reag. Ph. Eur.

agency

-

Quality Level

300

Quality Level

-

Quality Level

200

Quality Level

100

form

solid

form

crystalline, solid

form

powder or crystals

form

-


Still not finding the right product?

Explore all of our products under Boric acid


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
100165100004022536002606
100165500004022536002620
100165901204027269677657
100165010004022536002576
100165050004022536002583
100165000004022536002569
100165902504022536002644

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters