Skip to Content
Merck

Skip To

07-2192

Anti-phospho-TBK1 (Ser172) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Serine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer172)

Gene Information

human ... TBK1(29110)

General description

TBK, also known as IκB kinase (IKK)-related kinase NAK, belong to the serine/threonine IKK kinase family and is thought to play a role in NF-kappa-B (NF-κB) activation. The NF-κB complex of proteins is inhibited by I-kappa-B (IKB) proteins, which inactivate NF-κB by trapping it in the cytoplasm. Phosphorylation of serine residues on the IKB proteins by IKB kinases marks them for destruction via the ubiquitination pathway, thereby allowing activation and nuclear translocation of the NFKB complex. The protein encoded by this gene is similar to IKB kinases and can mediate NF-κB activation in response to certain growth factors. For example, the protein can form a complex with the IKB protein TANK and TRAF2 and release the NF-κB inhibition caused by TANK.
~90 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to human TBK1 phosphorylated at Ser172.

Application

This phospho-TBK1 antibody is validated for use in WB for the detection of the phospho-TBK1 protein.

Biochem/physiol Actions

This antibody recognizes TBK1 phosphorylated at Ser172.

Analysis Note

Control
EGF treated and untreated HEK293 cell lysates
Evaluated by Western Blot in EGF treated and untreated HEK293 cell lysates.

Western Blot Analysis: 1:1,000 dilution of this antibody detected TBK1 on 10 µg of EGF treated and untreated HEK293 cell lysates.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
06-1425ABS9204-736
species reactivity

mouse, human

species reactivity

human, mouse

species reactivity

human

species reactivity

mouse, human

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

SK703, monoclonal

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

conjugate

-

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

UniProt accession no.

Q9UHD2

UniProt accession no.

O43318

UniProt accession no.

Q13470

UniProt accession no.

P31749

Gene Information

human ... TBK1(29110)

Gene Information

human ... MAP3K7(6885)

Gene Information

human ... TNK1(8711)

Gene Information

human ... AKT1(207)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
07-219204053252292644

Questions

Reviews

No rating value

Active Filters