Skip to Content
Merck

Skip To

V-033

Tocopheryl acetate (Vitamin E acetate) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
UNSPSC Code:
41116107
EC Number:
200-835-2
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O1[C@@](CCc2c1c(c(c(c2C)OC(=O)C)C)C)(CCC[C@@H](CCC[C@@H](CCCC(C)C)C)C)C

InChI

1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1

InChI key

ZAKOWWREFLAJOT-CEFNRUSXSA-N

General description

Tocopheryl acetate is a natural form of Vitamin E, used as a fortifying agent and dietary supplement for Vitamin E in food. It is also the main ingredient of skin care products and dermatological preparations. Tocopheryl acetate in combination with α-tocopherol has been used in the therapy of conditions such as sterility, muscular dystrophy, cardiac vascular disorders and anemia.

Application

This internal standard is suitable for HPLC or LC-MS/MS Vitamin E testing applications such as quality control testing of Vitamin E fortified foods, dietary supplements, or pharmaceuticals.
Tocopheryl Acetate (Vitamin E acetate) may be used as an analytical standard for the determination of the analyte in aquatic organisms, fish feed and pharmaceutical formulations[1] by colorimetric and chromatography techniques, respectively.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
29992T3001T1157
technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

-

technique(s)

-

technique(s)

cell culture | insect: suitable

application(s)

clinical testing

application(s)

cell analysis
research use (small molecule)

application(s)

-

application(s)

-

form

liquid

form

liquid

form

oil or semi-solid

form

liquid (or semi-solid)

format

single component solution

format

-

format

-

format

-

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200

grade

certified reference material

grade

-

grade

-

grade

-


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Calorimetric and infrared spectroscopic studies of the interaction of a-tocopherol and a-tocopheryl acetate with phospholipid vesicles
Villalain J, et al.
European Journal of Biochemistry, 158(1), 141-147 (1986)
Simultaneous determination of alpha-tocopheryl acetate and tocopherols in aquatic organisms and fish feed
Huo JZ, et al
Journal of Chromatography. B, Biomedical Sciences and Applications, 724(2), 249-255 (1999)
A S Amin
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 51(3), 267-272 (2001-05-10)
A simple, accurate and sensitive method for the determination of micro amounts of vitamin E (V(E)) in pure form and in multi-vitamin capsules is described. The method is based on the reduction of tetrazolium blue in slightly alkaline medium by



Global Trade Item Number

SKUGTIN
V-033-1ML04061837428722

Questions

Reviews

No rating value

Active Filters