Skip to Content
Merck

Skip To

M-010

(±)-MDA-D5 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-3,4-Methylenedioxyamphetamine-d5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10D5H8NO2
CAS Number:
Molecular Weight:
184.25
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

NC(C([2H])([2H])[2H])([2H])C([2H])([H])C1=CC(OCO2)=C2C=C1

InChI

1S/C10H13NO2/c1-7(11)4-8-2-3-9-10(5-8)13-6-12-9/h2-3,5,7H,4,6,11H2,1H3/i1D3,4D,7D

InChI key

NGBBVGZWCFBOGO-KIUTZQKNSA-N

General description

A stable-labeled internal standard suitable for quantitation of MDA levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. MDA, or 3,4-methylenedioxyamphetamine, is an illicit recreational drug of the amphetamine and phenethylamine classes. Also known as tenamphetamine, MDA is abused for its psychedelic and hallucinogenic effects, similar to those of MDMA(Ecstasy).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M-027M-029M-011
application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under (±)-MDA-D5 solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
M-010-1ML04061833976081

Questions

Reviews

No rating value

Active Filters