Skip to Content
Merck

Skip To

L-028

Lisdexamfetamine-D4 dimesylate solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Lisdexamfetamine-D4 dimesylate solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H21D4N3O · C2H6S2O6H2
Molecular Weight:
459.61
NACRES:
NA.24
UNSPSC Code:
41116107
EC Number:
200-659-6
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CS(O)(=O)=O.CS(O)(=O)=O.[2H]C([2H])(CN)C([2H])([2H])C[C@H](N)C(=O)N[C@@H](C)Cc1ccccc1

InChI

1S/C15H25N3O.2CH4O3S/c1-12(11-13-7-3-2-4-8-13)18-15(19)14(17)9-5-6-10-16;2*1-5(2,3)4/h2-4,7-8,12,14H,5-6,9-11,16-17H2,1H3,(H,18,19);2*1H3,(H,2,3,4)/t12-,14-;;/m0../s1/i5D2,6D2;;

InChI key

CETWSOHVEGTIBR-DIDVIBMPSA-N

General description

A certified solution standard suitable for quantitation of lisdexamfetamine in urine or other biological samples by GC/MS or LC/MS in clinical toxicology testing, forensic analysis, urine drug testing, or pharmaceutical research. Lisdexamfetamine, a psychostimulant prodrug of dextroamphetamine, is marketed as the drug Vyvanse® for treatment of attention deficit hyperactivity disorder (ADHD) in adults and children six to twelve years of age.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vyvanse is a registered trademark of Shire, LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L-026M-185V-028
technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

format

single component solution

format

single component solution

format

single component solution

format

single component solution

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters