This product can be used for the application of trace analysis of EDDP in urine for in vitro tests that are specific to the presence of EDDP.
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1.0 mg/mL in methanol (as pyrrolinium)
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
O=Cl(=O)([O-])=O.C[N+]1=C(CC([2H])([2H])[2H])C(C2=CC=CC=C2)(C3=CC=CC=C3)CC1C
InChI
1S/C20H24N.ClHO4/c1-4-19-20(15-16(2)21(19)3,17-11-7-5-8-12-17)18-13-9-6-10-14-18;2-1(3,4)5/h5-14,16H,4,15H2,1-3H3;(H,2,3,4,5)/q+1;/p-1/i1D3;
InChI key
FICCBMGBBWWZRF-NIIDSAIPSA-M
General description
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable |
| format single component solution | format single component solution | format neat | format single component solution |
| form liquid | form liquid | form solid | form liquid |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Still not finding the right product?
Explore all of our products under EDDP-D3 perchlorate solution
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| E-062-1ML | 04061833592687 |
-
Is the the EDDPs from methadone use that they look for and can you can add to synthetic urine or urine to make it come up positive in a urine test
1 answer-
Helpful?
-





