Skip to Content
Merck

Skip To

C-015

Codeine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H21NO3
CAS Number:
Molecular Weight:
299.36
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
EC Number:
200-659-6
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal); Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

COc1ccc2C[C@@H]3[C@@H]4C=C[C@H](O)[C@@H]5Oc1c2[C@]45CCN3C

InChI

1S/C18H21NO3/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19/h3-6,11-13,17,20H,7-9H2,1-2H3/t11-,12+,13-,17-,18-/m0/s1

InChI key

OROGSEYTTFOCAN-DNJOTXNNSA-N

Gene Information

General description

Codeine is an opiate used for its analgesic, antitussive, and sedative properties and is the second-most predominant alkaloid in opium. This certified reference solution is suitable for use as a starting material in calibrators and controls for LC/MS and GC/MS testing methods in clinical toxicology, forensic analysis, pharmaceutical research, and pain prescription monitoring. As a widely popular recreational drug, codeine is regulated under various narcotic control laws in countries such as Australia, New Zealand, Romania, Sweden, and the United States. Since opiates are classified as controlled substances, Cerilliant offers most opiate reference standards as DEA-exempt solutions for added convenience.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-0061.090981.09910
application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

-

application(s)

-

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

titration: suitable

technique(s)

-

form

liquid

form

liquid

form

liquid

form

-

Quality Level

300

Quality Level

300

Quality Level

200

Quality Level

100

grade

certified reference material

grade

certified reference material

grade

-

grade

-

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

15-25°C

storage temp.

20-25°C


Still not finding the right product?

Explore all of our products under Codeine solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters