Skip to Content
Merck

Skip To

B-008

Benzoylecgonine-d3 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Benzoylecgonine-N-methyl-d3 solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16D3H16NO4
CAS Number:
Molecular Weight:
292.34
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

mass shift

M+3

shipped in

wet ice

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])N1[C@H]2CC[C@@H]1[C@H]([C@H](C2)OC(=O)c3ccccc3)C(O)=O

InChI

1S/C16H19NO4/c1-17-11-7-8-12(17)14(15(18)19)13(9-11)21-16(20)10-5-3-2-4-6-10/h2-6,11-14H,7-9H2,1H3,(H,18,19)/t11-,12+,13-,14+/m0/s1/i1D3

InChI key

GVGYEFKIHJTNQZ-GZBWLTEMSA-N

General description

A stable-labeled internal standard suitable for isotope dilution methods or benzoylecgonine testing in urine, saliva, blood, or hair by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing, pain prescription monitoring or forensic analysis. Benzoylecgonine is the primary metabolite of cocaine and a major biomarker for determination of cocaine use.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B-001B-014N-082
technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under Benzoylecgonine-d3 solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters