Skip to Content
Merck

Skip To

733792

Triphenyl phosphate solution

suitable for NMR (reference standard), 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(C6H5O)3PO
Molecular Weight:
326.28
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12190000
MDL number:
Form:
solution
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

concentration

0.0485 M in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

10 mm × 8 in.

suitability

suitable for NMR (reference standard)

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Features and Benefits

31P sensitivity

Preparation Note

10 mm O.D. tube contains 3.175 mL.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
708968733741733776
technique(s)

NMR: suitable

technique(s)

NMR: suitable

technique(s)

NMR: suitable

technique(s)

NMR: suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

solution

form

solution

form

solution

form

solution

concentration

0.0485 M in acetone-d6 (99.9 atom % D)

concentration

0.0485 M in acetone-d6 (99.9 atom % D)

concentration

0.0485 M in acetone-d6 (99.9 atom % D)

concentration

1% in acetone-d6 (99.9 atom % D)

suitability

suitable for NMR (reference standard)

suitability

suitable for NMR (reference standard)

suitability

suitable for NMR (reference standard)

suitability

suitable for NMR (reference standard)

NMR tube size

10 mm × 8 in.

NMR tube size

5 mm × 7 in. , 5 mm × 8 in.

NMR tube size

3 mm × 8 in.

NMR tube size

10 mm × 8 in.


Still not finding the right product?

Explore all of our products under Triphenyl phosphate solution


pictograms

FlameExclamation mark

signalword

Danger

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

-2.2 °F

flash_point_c

-19 °C

Hazard Classifications

Aquatic Chronic 3 - ED HH 1 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters