Skip to Content
Merck

Skip To

1367005

USP

Lincomycin hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Lincomycin hydrochloride monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H34N2O6S · HCl · H2O
CAS Number:
Molecular Weight:
461.01
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

lincomycin

manufacturer/tradename

USP

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].[H]O[H].CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O

InChI

1S/C18H34N2O6S.ClH.H2O/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H;1H2/t9-,10-,11+,12-,13+,14-,15-,16-,18-;;/m1../s1

InChI key

LFZGYTBWUHCAKF-DCNJEFSFSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lincomycin hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L0650000BP215L2774
application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

-

API family

lincomycin

API family

lincomycin

API family

lincomycin

API family

-

format

neat

format

neat

format

neat

format

-


Still not finding the right product?

Explore all of our products under Lincomycin hydrochloride


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Reza Tabrizi et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 72(5), 850-857 (2014-02-25)
The aim of the present study was to evaluate the periodontal regenerative capacity of demineralized freeze-dried bone allograft (DFDBA) alone or used with local lincomycin. In the present single-blind, randomized, controlled clinical trial, 20 subjects 26 years old or older
Sonja Hillen et al.
Veterinary microbiology, 172(1-2), 223-229 (2014-06-21)
Brachyspira hyodysenteriae, the causative agent of swine dysentery, is responsible for severe mucohaemorrhagic colitis with considerable financial loss to worldwide swine production. Antimicrobial resistance against macrolides and lincosamides is widespread and the mechanisms are well known. Currently, the most common
Biochemistry: Elusive source of sulfur unravelled.
Charles E Melançon
Nature, 518(7537), 45-46 (2015-01-22)



Global Trade Item Number

SKUGTIN
1367005-200MG04061838731388

Questions

Reviews

No rating value

Active Filters