Skip to Content
Merck

Skip To

SML1908

Syrosingopine

monocarboxylate lactate transporters 1 and 4 inhibitor, powder

Synonym(s):

(3β,16β,17α,18β,20α)-18-[[4-[(Ethoxycarbonyl)oxy]-3,5-dimethoxybenzoyl]oxy]-11,17-dimethoxy-yohimban-16-carboxylic acid methyl ester, Methyl reserpate ester of syringic acid ethyl carbonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C35H42N2O11
CAS Number:
Molecular Weight:
666.71
UNSPSC Code:
12352200
NACRES:
NA.77
Form:
powder
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Syrosingopine,

form

powder

Quality Level

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

SMILES string

O=C(OC)[C@@H]1[C@]2([H])C[C@@]3([H])C4=C(C5=CC=C(OC)C=C5N4)CCN3C[C@]2([H])C[C@H](OC(C6=CC(OC)=C(OC(OCC)=O)C(OC)=C6)=O)[C@H]1OC

InChI

1S/C35H42N2O11/c1-7-46-35(40)48-31-26(42-3)12-18(13-27(31)43-4)33(38)47-28-14-19-17-37-11-10-22-21-9-8-20(41-2)15-24(21)36-30(22)25(37)16-23(19)29(32(28)44-5)34(39)45-6/h8-9,12-13,15,19,23,25,28-29,32,36H,7,10-11,14,16-17H2,1-6H3/t19-,23+,25-,28-,29+,32+/m1/s1

InChI key

ZCDNRPPFBQDQHR-SSYATKPKSA-N

Application

Syrosingopine has been used as a monocarboxylate transporter (MCT) inhibitor
  • to study its effects on anti-CD147-induced metabolon disruption in human breast cancer cells[1]
  • to study its effects on Cryptosporidium parvum-infected HCT-8 cells[2]
  • in orthogonal linear separation analysis (OLSA)-derived decomposed analysis[3]

Biochem/physiol Actions

Syrosingopine is a derivative of reserpine[3] and inhibits monocarboxylate lactate transporters 1 and 4 (MCT1/4).[2]
Syrosingopine is an antihypertensive agent related to reserpine that was found to potentiate the anticancer effects of the antidiabetic agent metformin and phenformin without harmful effects on normal cells.
Syrosingopine is an antihypertensive agent related to reserpine that was found to potentiate the anticancer effects of the antidiabetic agent metformin and phenformin without harmful effects on normal cells. Syrosingopine was also found to potentiate the anticancer activity of mitochondrial electron transport chain (ETC) inhibitors. Its mechanism of action is currently unknown but may involve inhibition of glycolytic enzyme α-enolase rather than its known activity as an inhibitor of vesicular monoamine transporters VMAT1 and VMAT2.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0147R087583580
form

powder

form

powder

form

-

form

solid

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

-

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: ≥15

solubility

-

solubility

-

color

white to light brown

color

white to tan

color

-

color

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Shumpei Nemoto et al.
Journal of natural products, 84(4), 1283-1293 (2021-04-10)
It is difficult to understand the entire effect of a natural product because such products generally have multiple effects. We propose a strategy to understand these effects effectively by decomposing them with a profile data analysis method we developed. A
Don Benjamin et al.
Science advances, 2(12), e1601756-e1601756 (2016-12-29)
We report that the anticancer activity of the widely used diabetic drug metformin is strongly potentiated by syrosingopine. Synthetic lethality elicited by combining the two drugs is synergistic and specific to transformed cells. This effect is unrelated to syrosingopine's known
Juan Vélez et al.
Biology, 10(1) (2021-01-21)
Cryptosporidium parvum is an apicomplexan zoonotic parasite recognized as the second leading-cause of diarrhoea-induced mortality in children. In contrast to other apicomplexans, C.



Global Trade Item Number

SKUGTIN
SML1908-5MG04061832926681
SML1908-25MG04061832926674

Questions

Reviews

No rating value

Active Filters