This DNA ladder can be used with a loading buffer such as G2526:
https://www.sigmaaldrich.com/product/sigma/g2526
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.25
UNSPSC Code:
41141827
Biological source:
synthetic
Form:
liquid
Storage temp.:
−20°C
Pricing and availability is not currently available.
biological source
synthetic
Quality Segment
grade
Molecular Biology
form
liquid
usage
75 uses
suitability
suitable for electrophoresis (DNA)
storage temp.
−20°C
General description
Sigma′s PCR 100 bp Low Marker contains 10 dsDNA fragments from 100 – 1,000 bases, evenly spaced in 100 bp repeats. Two ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose gel.
The PCR 100 bp low ladder has been specially designed for the size determination of polymerase chain reaction (PCR) generated DNA fragments.
Application
PCR 100 bp Low Ladder has been used:
- in agarose gel electrophoresis as a standard DNA marker to obtain DNA fragments of about 240 bp for polymerase chain reaction (PCR)-denaturing gradient gel electrophoresis (DGGE) analysis[1]
- to visualize DNA from enzyme digested PCR mixture
- to estimate the molecular weight of the PCR product fragments
Other Notes
Contains 10 bands, 100–1,000 bp in exact 100 bp spaced (ladder) recombinant repeats.
For optimal resolution, the recommended agarose gel concentration is 2.0%.
Sigma′s PCR 100 bp Low Marker is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.
1 of 1
This Item | |||
|---|---|---|---|
| grade Molecular Biology | grade Molecular Biology | grade - | grade Molecular Biology |
| biological source synthetic | biological source - | biological source - | biological source fermentation/recombinant (E. coli strain DH5A) |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
| form liquid | form liquid | form liquid | form liquid |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| usage 75 uses | usage 75 uses | usage sufficient for 50 uses | usage 80 uses |
Still not finding the right product?
Explore all of our products under PCR 100 bp Low Ladder
Storage Class
10 - Combustible liquids
wgk
WGK 3
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
flash_point_f
No data available
flash_point_c
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P1473-1VL | 04061835516230 |
-
What loading buffer/dye is the P1473 DNA ladder compatible with for loading onto a gel.
1 answer-
Helpful?
-



