Skip to Content
Merck

Skip To

MINI26

PKH26 Red Fluorescent Cell Linker Mini Kit for General Cell Membrane Labeling

Distributed for Phanos Technologies

Synonym(s):

Red PKH membrane label kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 kit

Quality Level

manufacturer/tradename

Distributed for Phanos Technologies

storage condition

protect from light

technique(s)

flow cytometry: suitable

fluorescence

λex 551 nm; λem 567 nm (PKH26 dye)

application(s)

cell analysis
detection

detection method

fluorometric

shipped in

ambient

storage temp.

room temp

General description

This kit provides enough dye for up to 25 samples of 20 million cells each and enough diluent for up to 5 samples of 20 million cells each, when using a 2 mL final staining volume containing a final concentration of 2 x 10-6 M of PKH26 dye. Users must determine the optimal dye and cell concentrations from their cell type(s) and experimental purposes.

Application

PKH26 red fluorescent cell linker mini kit has been used for:
  • labelling microglial, natural killer cells and preadipocytes
  • tracking of viable transplanted adipose-derived stem cells
  • in vitro proliferation studies of bone marrow stem cells
  • general cell membrane labeling, including for in vitro cell labeling and long term in vivo cell tracking.

Other Notes

For additional technical details on PKH and CellVue® Fluorescent Cell Linker Dyes including an extensive bibliography, please visit here.

Legal Information

CellVue is a registered trademark of Phanos Technologies

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PKH26GLMINI67PKH26PCL
technique(s)

flow cytometry: suitable

technique(s)

-

technique(s)

flow cytometry: suitable

technique(s)

-

storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

storage condition

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

detection method

fluorometric

detection method

fluorometric

detection method

fluorometric

detection method

-

application(s)

cell analysis
detection

application(s)

-

application(s)

cell analysis
detection

application(s)

-

manufacturer/tradename

Distributed for Phanos Technologies

manufacturer/tradename

Distributed for Phanos Technologies

manufacturer/tradename

Distributed for Phanos Technologies

manufacturer/tradename

-


Kit Components Only

Product No.
Description

  • Diluent C 10 mL

  • PKH26 Cell Linker in ethanol .1 mL

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

57.2 °F - closed cup

flash_point_c

14 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Imaging of peripheral benzodiazepine receptor expression as biomarkers of detrimental versus beneficial glial responses in mouse models of Alzheimer's and other CNS pathologies
Ji B, et al.
The Journal of Neuroscience, 28(47), 12255-12267 (2008)
Maria Giovanna Gandolfi et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-04)
Exosomes derived from mesenchymal stem cells are extracellular vesicles released to facilitate cell communication and function. Recently, polylactic acid (PLA), calcium silicates (CaSi), and dicalcium phosphate dihydrate (DCPD) have been used to produce bioresorbable functional mineral-doped porous scaffolds-through thermally induced
Human leukocyte antigen-G1 inhibits natural killer cytotoxicity through blocking the activating signal transduction pathway and formation of activating immunologic synapse
Yu Y, et al.
Human immunology, 69(1), 16-23 (2008)



Global Trade Item Number

SKUGTIN
MINI26-1KT04061834066712

Questions

1–2 of 2 Questions  
  1. MINI26-1KTの実験プロトコルを入手したいのですが、どこにありますか?

    1 answer
    1. The protocol is published in the product datasheet, available in the 'DOCUMENTATION' section of this page, under the 'Mor Documents' tab. Please see the link below to review:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/271/475/mini26bul.pdf

      Helpful?

  2. Is it possible to lable cells with PKH26 at 4oC?

    1 answer
    1. The membrane dyes will remain stable during the monoclonal staining at 4 deg C; however, capping of the monoclonal antibodies is highly probable if the general cell membrane labeling is carried out at ambient temperature subsequent to antibody labeling. As additional information may be required to address the question completely, please navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquiries" under the Products Section with all the information needed so that a member of the Technical Service team can reach out to assist further.

      For more information, please refer to the product data-sheet below: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/271/475/mini26bul.pdf.

      Helpful?

Reviews

No rating value

Active Filters