Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
form
solution
Quality Level
shelf life
Expiry date on the label
IVD
for in vitro diagnostic use
concentration
10 % (w/v)
technique(s)
titration: suitable
application(s)
hematology
histology
storage temp.
room temp
SMILES string
O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O.OP(O)(O)=O
InChI
1S/2H3O4P.72O.24W/c2*1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h2*(H3,1,2,3,4);;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;
InChI key
XEFOGGWKYODFIU-UHFFFAOYSA-N
Application
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) titration: suitable | technique(s) - | technique(s) - | technique(s) - |
| concentration 10 % (w/v) | concentration 0.5 % (w/v) in water | concentration 0.5 % (w/v) in water | concentration 0.5 % (w/v) in water |
| form solution | form solution | form solution | form solution |
| application(s) hematology | application(s) hematology | application(s) hematology | application(s) hematology |
| shelf life Expiry date on the label | shelf life Expiry date on the label. | shelf life Expiry date on the label. | shelf life Expiry date on the label. |
| storage temp. room temp | storage temp. room temp | storage temp. room temp | storage temp. room temp |
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| HT152-250ML | 04061833835289 |


