Skip to Content
Merck

Skip To

FD70

Fluorescein isothiocyanate–dextran

average Mw 59,000-77,000

Synonym(s):

FITC–Dextran

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Leuconostoc mesenteroides)

Quality Level

conjugate

FITC conjugate

form

powder

mol wt

average Mw 59,000-77,000

extent of labeling

0.001-0.020 mol FITC per mol glucose

color

light yellow to dark orange

solubility

water: 25 mg/mL

storage temp.

2-8°C

General description

Dextran is a polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(166) linkages. The remaining (163) linkages account for the branching of dextran. Conflicting data on the branch lengths implies that the average branch length is less than three glucose units. However, other methods indicate branches of greater than 50 glucose units exist. Native dextran has been found to have a molecular weight (MW) in the range of 9 million to 500 million. Lower MW dextrans will exhibit slightly less branching and have a more narrow range of MW distribution. Dextrans with MW greater than 10,000 behave as if they are highly branched. As the MW increases, dextran molecules attain greater symmetry. Dextrans with MW of 2,000 to 10,000, exhibit the properties of an expandable coil. At MW below 2,000, dextran is more rod-like. The MW of dextran is measured by one or more of the following methods: low angle laser light scattering, size exclusion chromatography, copper-complexation and anthrone reagent colorometric reducing-end sugar determination and viscosity.

Application

Dextran labeled with fluorescein isothiocyanate for possible use in perfusion studies in animals.
FITC-dextran is used extensively in microcirculation and cell permeability research utilizing microfluorimetry. FITC-dextran has been used to study plant cell wall porosity and capillary permeability. Plasma proteins have been shown not to bind to FITC-dextran.
FITC-dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.[1] Small FITC-dextrans are also used to measure processes such as endocytosis and cell junction permeability. FITC-dextran 70kDa (FD-70) may be used as a model compound for development of methods to deliver molecules into protoplasts and to study drug release from structures such as hydrogels and liposomes. It may be used in microvascular and cell monolayer permeability studies.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Sigma typically tests the solubility of FITC dextrans in water at concentrations at or above 25 mg/ml. Solutions should be protected from light. In vivo, FITC-dextran is stable for more than 24 hours.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
90718FD40FD20
biological source

bacterial (Leuconostoc mesenteroides)

biological source

synthetic

biological source

bacterial (Leuconostoc mesenteroides)

biological source

bacterial (Leuconostoc mesenteroides)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

solubility

water: 25 mg/mL

solubility

-

solubility

H2O: 25 mg/mL

solubility

water: 25 mg/mL

form

powder

form

powder or crystals

form

powder

form

powder

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C

color

light yellow to dark orange

color

-

color

light yellow to dark orange

color

light yellow to dark orange


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Mahmoud Soliman et al.
Scientific reports, 8(1), 13931-13931 (2018-09-19)
Intestinal epithelial tight junctions (TJ) are a major barrier restricting the entry of various harmful factors including pathogens; however, they also represent an important entry portal for pathogens. Although the rotavirus-induced early disruption of TJ integrity and targeting of TJ
K Tsuchiya et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 28(12), 1792-1805 (2016-06-11)
Stimulation of transient receptor potential ankyrin 1 (TRPA1), which abundantly expressed in enterochromaffin cells (ECC), has been reported to exert apparently contradictory results in in vitro contractility and in vivo gastrointestinal (GI) transit evaluations. The pharmaceutical-grade Japanese traditional medicine daikenchuto
Jennifer Hirst et al.
eLife, 3, e02866-e02866 (2014-05-29)
The heterotetrameric AP and F-COPI complexes help to define the cellular map of modern eukaryotes. To search for related machinery, we developed a structure-based bioinformatics tool, and identified the core subunits of TSET, a 'missing link' between the APs and



Global Trade Item Number

SKUGTIN
FD70-250MG04061833033500
FD70-1G04061826682623
FD70-100MG04061833622551

Questions

Reviews

No rating value

Active Filters