Skip to Content
Merck

Skip To

F8263

Iron(II) sulfate heptahydrate

greener alternative

suitable for plant cell culture, ≥99%

Synonym(s):

Ferrous sulfate heptahydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
NACRES:
NA.71
PubChem Substance ID:
UNSPSC Code:
12352207
EC Number:
231-753-5
MDL number:
Assay:
≥99%
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

14.6 mmHg ( 25 °C)

Quality Level

assay

≥99%

shelf life

2 yr

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

cell culture | plant: suitable

density

1.898 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

SURQXAFEQWPFPV-UHFFFAOYSA-L

General description

Iron(II) sulfate heptahydrate is used in cell culture applications generally bound to transferrin, citrate or other iron transport molecules.
We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives. IRON(II) SULFATE HEPTAHYDRATE is essential for developing greener energy solutions, particularly in iron-based lithium-ion batteries. Its eco-friendly properties enhance energy storage efficiency, reduce reliance on harmful materials, and support the advancement of sustainable technologies. Click here for more information.

Application


  • Development of spherical iron(II) sulfate heptahydrate-containing solid particles with sustained drug release.: This study presents the development of spherical solid particles containing iron(II) sulfate heptahydrate aimed at sustained drug release applications. The focus was on optimizing the formulation for controlled release, which could be significant for future biochemistry and pharmacology research (Szabo-Revesz et al., 2007).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
9356892154221.03963
assay

≥99%

assay

99.99% trace metals basis

assay

≥99.0%

assay

-

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

500

vapor pressure

14.6 mmHg ( 25 °C)

vapor pressure

-

vapor pressure

14.6 mmHg ( 25 °C)

vapor pressure

-

density

1.898 g/mL at 25 °C (lit.)

density

-

density

1.898 g/mL at 25 °C (lit.)

density

1.89 g/cm3 at 20 °C

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

-

greener alternative category

Enabling

greener alternative category

Enabling

greener alternative category

Aligned

greener alternative category

-


Still not finding the right product?

Explore all of our products under Iron(II) sulfate heptahydrate


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yanyan Wei et al.
Food chemistry, 138(2-3), 1952-1958 (2013-02-16)
The present study evaluated the effectiveness of germination and iron fortification on iron concentration and bioavailability of brown rice. Iron fortification during germination process with 0.05-2 g/L ferrous sulfate increased the iron concentration in germinated brown rice from 1.1 to
Hsiao-Chi Chuang et al.
Toxicology and applied pharmacology, 266(3), 329-334 (2012-12-25)
Pulmonary epithelial lining fluid (ELF) is the first substance to make contact with inhaled particulate matter (PM) and interacts chemically with PM components. The objective of this study was to determine the role of ELF in oxidative stress, DNA damage
Xiao-Kui Ma et al.
Carbohydrate polymers, 92(2), 1188-1196 (2013-02-13)
The influence of several components on exopolysaccharide (EPS) production and antioxidative activity (TEAC, Trolox-Equivalent Antioxidant Capacity) as well as their effects on the morphological development and cell viability of Phellinus sp. P0988 was determined. Rutin, FeSO(4), Tween 80 and complex



Global Trade Item Number

SKUGTIN
F8263-1KG04061833620427
F8263-500G04061833620434

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters