Skip to Content
Merck

Skip To

F3418

Monoclonal Anti-Cytokeratin, pan−FITC antibody produced in mouse

clone C-11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-Pan Cytokeratin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
FITC conjugate
Clone:
C-11, monoclonal
Application:
IF (d), IHC (p), WB
Citations:
48
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C-11, monoclonal

form

buffered aqueous solution

species reactivity

bovine, mouse, frog, human, kangaroo rat, rat

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:50 using PtK2 cells, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50 using human placenta, western blot: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Cytokeratins are the intermediate filaments of endocrine cells. These proteins belong to the intermediate filament (10 nm) superfamily of cytoskeletal proteins. Cytokeratins are the largest and most complex group of intermediate filaments. The family consists of at least 30 proteins with molecular weights ranging from 40kDa to 68kDa.

Immunogen

keratin-enriched preparation from cultured human epidermoid carcinoma cell line A431.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Immunohistochemistry (1 paper)
Monoclonal Anti-Cytokeratin, pan−FITC antibody has been used in immunocytochemistry and has also been used to visualize lung epithelial cells.

Biochem/physiol Actions

Cytokeratins helps in subtyping several kinds of epithelial differentiation or for narrowing possible sites of origin for metastatic carcinomas. Cytokeratins may play an important role in transhepatic transport and canalicular secretion.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2931MAB3412SAB4700667
antibody form

purified immunoglobulin

antibody form

ascites fluid

antibody form

purified antibody

antibody form

purified immunoglobulin

species reactivity

bovine, mouse, frog, human, kangaroo rat, rat

species reactivity

bovine, mouse, frog, human, kangaroo rat, rat

species reactivity

chicken, mouse, monkey, human, rabbit, bovine, rat

species reactivity

mammals

clone

C-11, monoclonal

clone

C-11, monoclonal

clone

AE1/AE3, monoclonal

clone

C-11, monoclonal

Gene Information

bovine ... Krt1(100301161)
human ... KRT1(3848), KRT13(3858), KRT13(3858), KRT18(3875), KRT18(3875), KRT4(3851), KRT4(3851), KRT5(3852), KRT5(3852), KRT6A(3868), KRT6A(3868), KRT6B(3854), KRT6B(3854), KRT8(3856), KRT8(3856)
mouse ... Krt1(16678), Krt10(16661), Krt10(16661), Krt13(16663), Krt13(16663), Krt18(16668), Krt18(16668), Krt4(16682), Krt4(16682), Krt5(110308), Krt5(110308), Krt6a(16687), Krt6a(16687), Krt6b(16688), Krt6b(16688), Krt8(16691), Krt8(16691)
rat ... Krt1(300250), Krt1-18(706059), Krt1-18(706059), Krt10(450225), Krt10(450225), Krt2-5(369017), Krt2-5(369017), Krt2-8(25626), Krt2-8(25626)

Gene Information

bovine ... Krt1(100301161)
human ... KRT1(3848), KRT13(3858), KRT13(3858), KRT18(3875), KRT18(3875), KRT4(3851), KRT4(3851), KRT5(3852), KRT5(3852), KRT6A(3868), KRT6A(3868), KRT6B(3854), KRT6B(3854), KRT8(3856), KRT8(3856)
mouse ... Krt1(16678), Krt10(16661), Krt10(16661), Krt13(16663), Krt13(16663), Krt18(16668), Krt18(16668), Krt4(16682), Krt4(16682), Krt5(110308), Krt5(110308), Krt6a(16687), Krt6a(16687), Krt6b(16688), Krt6b(16688), Krt8(16691), Krt8(16691)
rat ... Krt1(300250), Krt1-18(706059), Krt1-18(706059), Krt10(450225), Krt10(450225), Krt2-5(369017), Krt2-5(369017), Krt2-8(25626), Krt2-8(25626)

Gene Information

bovine ... Krt1(100301161)
human ... KRT1(3848)
mouse ... Krt1(16678)
rat ... Krt1(300250)

Gene Information

human ... KRT1(3848)

conjugate

FITC conjugate

conjugate

unconjugated

conjugate

-

conjugate

Allophycocyanin conjugate

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


Matthew L Tomlinson et al.
Frontiers in nutrition, 4, 61-61 (2018-01-13)
Flavonoids are a diverse group of plant secondary metabolites, known to reduce inflammatory bowel disease symptoms. How they achieve this is largely unknown. Our study focuses on the gut epithelium as it receives high topological doses of dietary constituents, maintains
C J Morrow et al.
Annals of oncology : official journal of the European Society for Medical Oncology, 27(6), 1155-1160 (2016-03-26)
Over the past decade, numerous reports describe the generation and increasing utility of non-small-cell lung cancer (NSCLC) patient-derived xenografts (PDX) from tissue biopsies. While PDX have proven useful for genetic profiling and preclinical drug testing, the requirement of a tissue
Ingunn M Stromnes et al.
Cancer immunology research, 5(11), 978-991 (2017-10-27)
Pancreatic ductal adenocarcinoma (PDA) is a lethal malignancy resistant to most therapies, including immune checkpoint blockade. To elucidate mechanisms of immunotherapy resistance, we assessed immune parameters in resected human PDA. We demonstrate significant interpatient variability in T-cell number, localization, and



Global Trade Item Number

SKUGTIN
F3418-.5ML04061838163424
F3418-.2ML04061838105226

Questions

Reviews

No rating value

Active Filters