Skip to Content
Merck

Skip To

F0504

Folate binding protein from bovine milk

lyophilized powder

Synonym(s):

Folate Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.61
MDL number:
Form:
lyophilized powder
Biological source:
bovine milk
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine milk

Quality Segment

form

lyophilized powder

usage

 mg sufficient for ~4,000 assays

composition

Protein, 20-40% modified Warburg-Christian

technique(s)

ligand binding assay: suitable, surface plasmon resonance (SPR): suitable

capacity

1.5-6.0 μg/mg, protein binding capacity (folic acid)(25 °C at pH 7.5.)

storage temp.

−20°C

Application

Folate binding protein can be used in radioligand binding assays of serum folate. The folate receptor has been used in studies as a therapeutic target in a variety of cancers.
Useful for radioligand binding assays of serum folate. May also bind N-methyltetrahydrofolic acid and other folate analogues.[1][2][3]
Folate binding protein from bovine milk has been used in folate binding studies using surface plasmon resonance and in the uptake studies in kidney proximal tubule cells.

Biochem/physiol Actions

Folate-binding protein (FBP) is present as a cell receptor and binds to folate in equimolar ratio. It is involved in the transport of folate and improves their bioavailability.

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing buffer salt as sodium citrate

Analysis Note

Purified over 1,000 fold. May contain a trace of β-mercaptoethanol.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7030C8654P1431
technique(s)

ligand binding assay: suitable, surface plasmon resonance (SPR): suitable

technique(s)

ELISA: suitable, blood typing: suitable, ligand binding assay: suitable

technique(s)

electrophoresis: suitable, immunocytochemistry: suitable, ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

biological source

bovine milk

biological source

bovine

biological source

bovine milk

biological source

bovine testis

form

lyophilized powder

form

lyophilized powder

form

powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

capacity

1.5-6.0 μg/mg, protein binding capacity (folic acid)(25 °C at pH 7.5.)

capacity

-

capacity

-

capacity

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters