Skip to Content
Merck

Skip To

D7906

Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide

chiral derivatizing agent , powder

Synonym(s):

FDAA, Marfey’s reagent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H9FN4O5
CAS Number:
Molecular Weight:
272.19
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.25
Beilstein/REAXYS Number:
6820069
MDL number:
Assay:
≥98% (TLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, powder

assay

≥98% (TLC)

Quality Level

form

powder

color

, Yellow to Light Yellow with Green

solubility

acetone: 10 mg/mL, clear (Yellow to Light Yellow with Green)

storage temp.

2-8°C

SMILES string

C[C@H](Nc1cc(F)c(cc1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

InChI

1S/C9H9FN4O5/c1-4(9(11)15)12-6-2-5(10)7(13(16)17)3-8(6)14(18)19/h2-4,12H,1H3,(H2,11,15)/t4-/m0/s1

InChI key

NEPLBHLFDJOJGP-BYPYZUCNSA-N

Application

N-α-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide is suitable for use for the derivatization of amino acids. glutamate and analine present in cell wall. Derivatized D- and L-amino acids can be resolved and quantitated by HPLC.

Biochem/physiol Actions

N-α-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide (FDAA) is a chiral derivatizing agent and is used routinely to improve the detection of underivatized amino acids in high performance liquid chromatography. Its usage is effective in separating stereoisomer.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7147820425384216
assay

≥98% (TLC)

assay

≥99.0% (sum of enantiomers, TLC)

assay

-

assay

-

solubility

acetone: 10 mg/mL, clear (Yellow to Light Yellow with Green)

solubility

-

solubility

-

solubility

-

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

form

powder

form

powder

form

-

form

liquid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

color

, Yellow to Light Yellow with Green

color

-

color

-

color

-


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



C3 and 2D C3 Marfey?s methods for amino acid analysis in natural products
Vijayasarathy S, et al.
Journal of Natural Products, 79(2), 421-427 (2016)
Purification of branched-chain amino acid aminotransferase from Helicobacter pylori NCTC 11637
Saito M, et al.
Amino Acids, 33(3), 445-449 (2007)
The functional dlt operon of Clostridium butyricum controls the D-alanylation of cell wall components and influences cell septation and vancomycin-induced lysis
Wydau-Dematteis S, et al.
Anaerobe, 35, 105-114 (2015)



Global Trade Item Number

SKUGTIN
D7906-100MG04061835354528
D7906-25MG04061835354535

Questions

Reviews

No rating value

Active Filters