Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
≥250 units/μg protein
Recombinant:
expressed in Sf9 cells
Pricing and availability is not currently available.
recombinant
expressed in Sf9 cells
Quality Segment
form
solution
specific activity
≥250 units/μg protein
shipped in
dry ice
storage temp.
−70°C
General description
Full length sequence with an N-terminal GST tag
Application
Biochem/physiol Actions
DPP IX is a member of the S9b family of dipeptidyl peptidases and cleaves the peptide hormones glucagon-like peptide-1, glucagon-like peptide-2, neuropeptide Y and peptide YY with marked kinetic differences compared with dipeptidyl peptidase IV. It has a role in regulating cell survival and proliferation through modulating molecular signaling cascades. Protein kinase B activation is significantly inhibited by human DPP9 overexpression in human hepatoma cells (HepG2 and Huh7) and human embryonic kidney cells (HEK293T)[1]. DPP9 is thought to influence cell-extracellular matrix interactions and may be involved in the regulation of tissue remodeling [2].
Physical form
Supplied as a solution in 40 mM Tris-HCl pH 8.0, 110 mM NaCl, 2.2 mM KCl, 20% glycerol, 3 mM DTT, and 16 mM glutathione
Other Notes
DPP IX is a member of the S9b family of dipeptidyl peptidases. and cleaves the peptide hormones glucagon-like peptide-1, glucagon-like peptide-2, neuropeptide Y and peptide YY with marked kinetic differences compared with dipeptidyl peptidase IV.
One unit will hydrolyze 1.0 picomole of H-Gly-Pro-AMC per minute at pH 7.5 at 25 deg °C
1 of 1
This Item | |||
|---|---|---|---|
| specific activity ≥250 units/μg protein | specific activity ≥20 units/μg protein | specific activity ≥1,500 units/μg protein, ≥1500 units/μg protein | specific activity ≥400 units/μg protein |
| form solution | form solution | form solution | form solution |
| recombinant expressed in Sf9 cells | recombinant expressed in Sf9 cells | recombinant expressed in Sf9 cells | recombinant expressed in Sf9 cells |
| storage temp. −70°C | storage temp. −70°C | storage temp. −70°C | storage temp. −70°C |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Storage Class
10 - Combustible liquids
wgk
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| D3071-10UG | 04061832700472 |



