Skip to Content
Merck

Skip To

CCF100

CM Sepharose

Fast Flow

Synonym(s):

Carboxymethyl Sepharose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
47101511
NACRES:
NA.56
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

suspension

technique(s)

affinity chromatography: suitable

matrix

6% cross-linked agarose

particle size

45-165 μm (wet bead)

pore size

4,000,000 exclusion limit (av. mol. wt.)

pH

4—13

capacity

90-130 μeq/mL(gel basis)

storage temp.

2-8°C

General description

Preswollen in 20% ethanol
CCF100-500ml′s updated product number is GE17-0719-01
25 ml is GE17-0719-10

Application

CM Sepharose is used in affinity chromatography, protein chromatography and ion exchange chromatography. CM Sepharose hase been used to study diet supplements and to characterize biochemical and enzymatic properties of Viperidae venom.

Legal Information

Sepharose is a trademark of Cytiva

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DFF100P7892O6001
matrix

6% cross-linked agarose

matrix

6% cross-linked agarose

matrix

agarose, 4% cross-linked

matrix

agarose, 4% cross-linked

particle size

45-165 μm (wet bead)

particle size

-

particle size

45-165 μm

particle size

45-165 μm

pore size

4,000,000 exclusion limit (av. mol. wt.)

pore size

~4,000,000 Da exclusion limit

pore size

-

pore size

-

form

suspension

form

suspension

form

suspension

form

-

capacity

90-130 μeq/mL(gel basis)

capacity

110-160 μeq/mL binding capacity(gel volume)

capacity

15-20 mg/mL binding capacity (human serum albumin), 3-5 mg/mL binding capacity (β-lactoglobulin)

capacity

3-5 mg/mL binding capacity (β-lactoglobulin)

technique(s)

affinity chromatography: suitable

technique(s)

affinity chromatography: suitable

technique(s)

-

technique(s)

-


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

100.4 - 109.4 °F

flash_point_c

38 - 43 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters