Skip to Content
Merck

Skip To

C8273

Cytochalasin D

From Zygosporium mansonii, ≥98% (TLC and HPLC), actin polymerization inhibitor, powder

Cytochalasin D

Synonym(s):

Lygosporin A, Zygosporin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C30H37NO6
CAS Number:
Molecular Weight:
507.62
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
244-804-1
MDL number:
Beilstein/REAXYS Number:
1632828
Assay:
≥98% (TLC and HPLC)
Form:
powder
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder

biological source

Zygosporium mansonii

Quality Level

assay

≥98% (TLC and HPLC)

form

powder

solubility

DMSO: soluble, ethanol: soluble

antibiotic activity spectrum

fungi

mode of action

DNA synthesis | interferes

storage temp.

−20°C

SMILES string

[H][C@@]12[C@H](C)C(=C)[C@@H](O)[C@@H]3\C=C\C[C@H](C)C(=O)[C@](C)(O)\C=C\[C@@H](OC(C)=O)[C@@]13C(=O)N[C@H]2Cc4ccccc4

InChI

1S/C30H37NO6/c1-17-10-9-13-22-26(33)19(3)18(2)25-23(16-21-11-7-6-8-12-21)31-28(35)30(22,25)24(37-20(4)32)14-15-29(5,36)27(17)34/h6-9,11-15,17-18,22-26,33,36H,3,10,16H2,1-2,4-5H3,(H,31,35)/b13-9+,15-14+/t17-,18+,22-,23-,24+,25-,26+,29+,30+/m0/s1

InChI key

SDZRWUKZFQQKKV-JHADDHBZSA-N

General description

Cytochalasin D is a cell-permeable fungal toxin. It disrupts actin microfilaments and activates the p53-dependent pathways causing arrest of the cell cycle at the G1-S transition. Cytochalasin D inhibits smooth muscle contraction and insulin-stimulated but not basal transport of glucose.
Cytochalasin D (CytoD) is commonly used to inhibit actin dynamics and study its role in cellular processes. This fungal-derived small organic compound acts as an inhibitor of the interaction between G-actin and cofilin by binding to G-actin. Additionally, CytoD inhibits the binding of cofilin to F-actin and reduces the rate of actin polymerization and depolymerization in living cells.

Application

Cytochalasin D has been used:
  • to treat H9c2 cells to study the relationship between cell stiffness and the F-actin cytoskeleton
  • as inhibitor of receptor-mediated endocytosis to treat oligodendrocyte precursor cells (OPCs) to investigate the effect of inhibiting specific extracellular vesicle (EV) uptake pathways
  • in in vitro embryo culture to artificially activate oocytes

Biochem/physiol Actions

Potent inhibitor of actin polymerization; disrupts actin microfilaments; activates the p53-dependent pathways; inhibits smooth muscle contraction; inhibits insulin-stimulated glucose transport.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7698C2618208851
assay

≥98% (TLC and HPLC)

assay

≥94% (TLC)

assay

-

assay

≥98% (HPLC)

form

powder

form

powder

form

ready-to-use solution

form

crystalline (powder)

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: soluble, ethanol: soluble

solubility

ethanol: soluble, clear to hazy

solubility

-

solubility

ethanol: 2 mg/mL, DMSO: soluble

mode of action

DNA synthesis | interferes

mode of action

protein synthesis | interferes

mode of action

enzyme | interferes

mode of action

-


Still not finding the right product?

Explore all of our products under Cytochalasin D


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Cytochalasin D acts as an inhibitor of the actin-cofilin interaction
Shoji K, et al.
Biochemical and Biophysical Research Communications (2012)
Advances of xenogeneic ovarian extracellular matrix hydrogels for in vitro follicle development and oocyte maturation
Frances-Herrero E, et al.
Biomaterials advances (2023)
Extracellular vesicle-associated cholesterol supports the regenerative functions of macrophages in the brain
Vanherle S, et al.
Journal of Extracellular Vesicles (2023)



Global Trade Item Number

SKUGTIN
C8273-10MG04061833522042
C8273-5MG04061833522066
C8273-1MG04061833522059

Questions

1–2 of 2 Questions  
  1. Could you provide the maximum solubility in DMSO (mg/mL)?

    1 answer
    1. This material is soluble in DMSO up to 371 mg/mL. Stock solutions prepared in dimethyl sulfoxide and stored at 22–24 °C have been reported to retain tissue culture activity for up to three months.

      Helpful?

  2. What is the time frame for the effect to take place in the culture media for all three products? How long will the effect last? What is the duration for which the cell can survive or tolerate the effect?

    1 answer
    1. The time frame for the effect to take place in the culture media, the duration of the effect, and the cell's ability to survive or tolerate the effect can be found in the following articles:

      How long does it take for the product to take effect in the culture media?

      J Cell Biol. 1974 May;61(2):481-500. doi: 10.1083/jcb.61.2.481.
      Action of cytochalasin D on cells of established lines. I. Early events
      PMID: 4208074

      How long will the effect last?

      Cell Motil Cytoskeleton. 1989;13(3):127-44. doi: 10.1002/cm.970130302.
      Latrunculins--novel marine macrolides that disrupt microfilament organization and affect cell growth: I. Comparison with cytochalasin D
      PMID: 2776221

      How long can the cell survive/tolerate?

      J Biol Chem. 1999 Feb 12;274(7):4259-65. doi: 10.1074/jbc.274.7.4259.
      Actin stabilization by jasplakinolide enhances apoptosis induced by cytokine deprivation
      PMID: 9933626

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review