Skip to Content
Merck

Skip To

C7352

L-Cysteine

from non-animal source, BioReagent, suitable for cell culture, ≥98%

Synonym(s):

(R)-2-Amino-3-mercaptopropionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HSCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
121.16
NACRES:
NA.75
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-158-2
MDL number:
Beilstein/REAXYS Number:
1721408
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source

Quality Segment

product line

BioReagent

assay

≥98%

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

240 °C (dec.) (lit.)

solubility

H2O: 25 mg/mL

SMILES string

N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m0/s1

InChI key

XUJNEKJLAYXESH-REOHCLBHSA-N

General description

L-Cysteine is one of the sulfur containing proteinogenic amino acid incorporated into proteins as directed by the genetic code. The thiol-side chain participates in a variety of oxidation/reduction reactions within the cell. The side chain participates in the formation of β bonds that modulate the secondary and ternary structure of proteins. L-Cysteine is an essential precursor for inorganic sulfates, coenzyme A. Cysteine is an essential and limiting for the formation of glutathione, an important antioxidant, within the cell. Mammalian liver can synthesize L-cysteine and is a semi-essential amino acid. L-cysteine prevents oxidation of nerve cells, aids in maintaining the stability and rigidity of connective tissues. L-cysteine speeds up the healing process of the body and reduces inflammatory reactions. L-cysteine protects testis partially against acrylamide induced toxicity. L-cysteine can prevent DNA damage during cryopreservation and preserves semen quality.
L-Cysteine is a proteinogenic amino acid incorporated into proteins as directed by the genetic code. The thiol-side chain participates in a variety of oxidation/reduction reactions within the cell. The side chain participates in the formation of β bonds that modulate the secondary and ternary structure of proteins. Cysteine is essential and limiting for the formation of glutathione, an important antioxidant, within the cell.

Application

L-Cysteine has been used:
  • as a media supplement in Trypticase-Yeast Extract-Iron-Serum (TYI-S-33) media for the culture of Giardia lamblia, to make enzyme solution in Dulbecco′s modified Eagles medium (DMEM) to establish hippocampus culture
  • to test its protective effect on acrylamide induced toxicity in testis
  • in the preparation of tris acetic acid extender solution for cryopreservation of semen
  • as an additive to make papain digestion solution

Biochem/physiol Actions

NMDA glutamatergic receptor agonist.

Other Notes

L-Cysteine is used to supplement cell culture media. This product is a non-animal source amino acid useful for serum-free media formulations intended for biomanufacturing applications.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
95437168149778672
form

crystalline powder

form

-

form

solid

form

powder

technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

assay

≥98%

assay

-

assay

97%

assay

≥98.0%

product line

BioReagent

product line

TraceCERT®

product line

-

product line

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

100

impurities

endotoxin, tested

impurities

-

impurities

-

impurities

≤0.5% ninhidrin positive substances (each), ≤10 ppm heavy metals, ≤300 ppm ammonium


Still not finding the right product?

Explore all of our products under L-Cysteine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. I have your product L-cysteine (C7352) powder. What is the solvent that I can use to resuspend the powder?

    1 answer
    1. As listed in the 'PROPERTIES' section, this product is soluble in water at 25 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters