Skip to Content
Merck
All Photos(1)

Documents

B3133

Sigma-Aldrich

Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride

trypsin substrate, chromogenic, ≥99% (TLC), powder, suitable for substrate for trypsin

Synonym(s):

L-BAPA, L-BAPNA, BANI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N6O4 · HCl
CAS Number:
Molecular Weight:
434.88
Beilstein:
4081878
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, ≥99% (TLC), suitable for substrate for trypsin

Assay

≥99% (TLC)

form

powder

solubility

acetone: water (1:1): 50 mg/mL

suitability

suitable for substrate for trypsin

storage temp.

−20°C

SMILES string

Cl[H].NC(=N)NCCC[C@H](NC(=O)c1ccccc1)C(=O)Nc2ccc(cc2)[N+]([O-])=O

InChI

1S/C19H22N6O4.ClH/c20-19(21)22-12-4-7-16(24-17(26)13-5-2-1-3-6-13)18(27)23-14-8-10-15(11-9-14)25(28)29;/h1-3,5-6,8-11,16H,4,7,12H2,(H,23,27)(H,24,26)(H4,20,21,22);1H/t16-;/m0./s1

InChI key

DEOKFPFLXFNAON-NTISSMGPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride is a substrate for trypsin.

Application

Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride has been used as a substrate:
  • for chymotrypsin and trypsin activity assay in enzymatic extract (EE) of M. tenellum and pancreatic tissue homogenates
  • in trypsin and papain standard curve generation
  • in serine protease and fibrinogenolytic assays of eupolytin1 protein

Linkage

Similar to B3279, but produced for Sigma.

Substrates

Chromogenic substrate for trypsin and other proteolytic enzymes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic Assays and Enzyme Histochemistry of Tuta absoluta Feeding on Tomato Leaves
Hamza R, et al.
BMC plant biology, 8(17), 1256-1263 (2018)
In vitro protein digestibility of animal, vegetal and microbial feed ingredients for Macrobrachium tenellum
Nolasco-Soria H, et al.
Latin American Journal of Aquatic Research, 46(3), 157-162 (2018)
Robert L Pizzey et al.
International dental journal, 61 Suppl 3, 33-40 (2011-11-02)
This study aimed to evaluate antimicrobial effects of an o-cymen-5-ol/zinc system. o-Cymen-5-ol and zinc gluconate minimum inhibitory concentration (MIC) and minimum bactericidal concentration (MBC) were determined against Streptococcus mutans, Actinomyces viscosus, Porphyromonas gingivalis, Fusobacterium nucleatum and Candida albicans. Synergy was
Feeding colostrum, its composition and feeding duration variably modify proliferation and morphology of the intestine and digestive enzyme activities of neonatal calves
Blattler U, et al.
The Journal of Nutrition, 131(4), 1256-1263 (2001)
The action of trypsin on synthetic chromogenic arginine substrates
Somorin O, et al.
Journal of Biochemistry, 85(1), 157-162 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service