Skip to Content
Merck

Skip To

A6778

Angiotensin Converting Enzyme from rabbit lung

≥2.0 units/mg protein (modified Warburg-Christian)

Synonym(s):

ACE, Peptidyl-dipeptidase A, Peptidyldipeptide Hydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
eCl@ss:
42010109
NACRES:
NA.32
EC Number:
MDL number:
Specific activity:
≥2.0 units/mg protein (modified Warburg-Christian)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

Quality Level

specific activity

≥2.0 units/mg protein (modified Warburg-Christian)

shipped in

dry ice

storage temp.

−20°C

General description

The angiotensin-converting enzyme (ACE) is a dipeptidyl-carboxypeptidase which exists in somatic and testicular isoforms with zinc binding motif HEXXH in their active site. ACE regulates blood pressure through renin-angiotensin system. ACE elevates blood pressure by converting angiotensin I to a key vasoconstrictor angiotensin II and inhibiting a potent vasodilator bradykinin. Inhibition of ACE is a targeted therapeutic strategy for high blood pressure. Several ACE synthetic inhibitory peptides available for clinical use include captopril, enalapril and lisinopril. Currently, developing inhibitory peptides from natural food sources, or phenolic compounds from plant sources to inhibit ACE is underway. ACE plays a critical role in fertilization by releasing the proteins anchored to glycosylphosphatidylinositol (GPI) in sperm membrane.

Application

Angiotensin converting enzyme from rabbit lung has been used:
  • for measuring inhibitory effect of egg white protein hydrolysates on ACE activity by high performance liquid chromatography (HPLC)
  • to measure the ACE inhibition by litchi pericarp and cooked chicken breast using hippuryl-L-histidyl-L-leucine (HHL) as substrate by reverse phase-HPLC (RP-HPLC)3 and HPLC respectively
  • in releasing GPI anchored protein in vitro in few cell lines like HeLa, HEK293 and in vivo in mice sperm.

Biochem/physiol Actions

Removes C-terminal dipeptides from susceptible substrates, e.g., angiotensin I and bradykinin.

Analysis Note

May contain traces of sodium chloride.

Other Notes

One unit will produce 1.0 μmole of hippuric acid from Hippuryl-His-Leu per min in 50 mM HEPES and 300 mM NaCl at pH 8.3 at 37 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2580SAE0075176872
specific activity

≥2.0 units/mg protein (modified Warburg-Christian)

specific activity

≥10 units/mg protein (Bradford)

specific activity

≥10 U/mg

specific activity

≥0.8 units/μg protein

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

liquid

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−70°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Angiotensin-converting enzyme is a GPI-anchored protein releasing factor crucial for fertilization
Kondoh G, et al.
Nature Medicine, 11(2), 160-166 (2005)
Isolation and identification of angiotensin-converting enzyme inhibitory peptides from egg white protein hydrolysates
Liu J, et al.
Food Chemistry, 122(4), 1159-1163 (2010)
Angiotensin converting enzyme (ACE) inhibitory peptides derived from the simulated in vitro gastrointestinal digestion of cooked chicken breast
Sangsawad P, et al.
Journal of functional foods, 29, 77-83 (2017)



Global Trade Item Number

SKUGTIN
A6778-.1UN04061835553525
A6778-1UN04061835540594
A6778-.25UN04061835553532
A6778-2UN04061835540600
A6778-5UN04061835511723

Questions

1–5 of 5 Questions  
  1. How many grams per 1 U?

    1 answer
    1. The milligram quantity of this product will vary from lot to lot. The vial content is dependent upon the protein content and enzymatic activity. The minimum protein content will be 70% and the minimum activity will be 2 units per milligram protein. As an example, a batch that reports a protein content of 100% and an activity of 2 units per milligram protein will have a vial content of 2.5 mg for the 5 unit package size. The protein content and activity are reported in the product Certificate of Analysis. Please see the link below to review a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/a6778#product-documentation

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. What is the effect of bleomycin on Angiotensin Converting Enzyme (ACE) activity in vivo?

    1 answer
    1. Bleomycin causes a marked reduction in rabbit lung ACE activity. Reference: Cancer Research, 40, 3621-3626, (1980).

      Helpful?

  4. Are there any other known inhibitors of rabbit lung Angiotensin Converting Enzyme (ACE), Product A6778?

    1 answer
    1. Idrapril, the prototype of a new class of angiotensin converting enzyme (ACE) inhibitors, competitively inhibits, with nanomolar apparent Ki, the hydrolysis of hippuryl-glycyl-glycine by rabbit lung ACE.  Kinetic Ki and kon and koff values were 470 pM, 3.0±1.5×106 M-1sec-1 and 1.4±0.3×10-3 sec-1, respectively.  Reference: Biochemical Pharmacology, 45(6), 1358-1362 (1993).

      Helpful?

  5. What is the medical significance of Angiotensin Converting Enzyme (ACE), Product A6778?

    1 answer
    1. Angiotensin-converting enzyme (ACE) produces angiotensin II, a substance that affects the cardiovascular system in numerous ways, including narrowing of blood vessels. This narrowing can cause high blood pressure and cause the heart to work harder.  ACE inhibitors treat a variety of conditions, such as high blood pressure, scleroderma and migraines. (Mayo Clinic)

      Helpful?

Reviews

No rating value

Active Filters