Skip to Content
Merck

Skip To

96096

Zeolite

Synonym(s):

Aluminosilicate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41123003
NACRES:
NA.25
EC Number:
215-283-8
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Level

particle size

<20 μm

SMILES string

[Si](=O)=O.[Al](=O)O[Al]=O

InChI

1S/2Al.O2Si.3O/c;;1-3-2;;;

InChI key

HNPSIPDUKPIQMN-UHFFFAOYSA-N

General description

Choosing a Molecular Sieve:
1. The preferential adsorption of one molecule over another depends upon pore diameter and mesh size.
2. The dynamic capacity of the silica gel for adsorbing a particular compound is governed by the internal surface area; the greater the surface area, the greater the dynamic capacity.
3. Rate of adsorption and sharpness of resolution are dependent chiefly on particle size and column packing; a fine particle size gives optimum sharpness of resolution.
4. Liquids are most readily adsorbed from solvents in which they are least soluble; a good solvent makes a good desorbing agent.
5. Highly polar liquids are readily adsorbed. Compounds having hydroxyl groups, or containing oxygen are strongly adsorbed.
6. For similar compounds, the higher the molecular weight the higher the selectivity.
7. For hydrocarbons of similar molecular weight, adsorptivity increases with the number of double bonds.
Regeneration or Activation:
A saturated molecular sieve can be restored to its original capacity by regeneration, the principle of which involves changing the conditions surrounding the adsorbent to correspond to a very low equilibrium capacity. In general, the greater the difference between the equilibrium capacities of adsorption and regeneration, the more rapid and complete the regeneration.
The sieve may be regenerated in one of four ways:
1. Thermal reactivation –The maximum regeneration temperature for Silica is 300°C.
2. Pressure reactivation
3. Passing an appropriate fluid through the gel bed at normal temperature and pressure.
4. Displacement of adsorbates by passing a high concentration of molecules in a fluid through the bed.

Application

Zeolite, a microporous, aluminosilicate mineral, is used as a molecular sieve and as an adsorbent of a wide variety of cations, such as Na+, K+, Ca2+, Mg2+ and others. Zeolite is used in the development of a variety of bioreactor devices.

Other Notes

In the determination of serum triglycerides.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
NIST8851NISTRM8850NIST8852
particle size

<20 μm

particle size

-

particle size

-

particle size

-

form

powder

form

solid

form

-

form

solid

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

-


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hossein Jahangirian et al.
International journal of nanomedicine, 15, 1005-1020 (2020-02-28)
The aim of this study was to prepare zeolite/iron (III) oxide nanocomposites (zeolite/Fe2O3-NCs) as a smart fertilizer to improve crop yield and soil productivity. Zeolite/Fe2O3-NCs were successfully produced by loading of Fe2O3-NPs onto the zeolite surface using a quick green
C Mery et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 47(3), 420-427 (2012-02-11)
An evaluation of natural zeolite as a microorganism carrier in nitrifying reactors operated in batch mode was carried out. Specifically, the influence of zeolite particle sizes of 0.5, 1.0 and 2.0 mm in diameter on microorganism adherence to zeolite, ammonium
Lactic acid production on liquid distillery stillage by Lactobacillus rhamnosus immobilized onto zeolite.
Djukic-Vukovic AP, Mojovic LV, Jokic BM, et al.
Bioresource Technology (2012)



Global Trade Item Number

SKUGTIN
96096-500G04061833317327
96096-100G04061833317303

Questions

1–10 of 17 Questions  
  1. Is it possible to encapsulate in it?

    1 answer
    1. Zeolite is reportedly often utilized to encapsulate metals for use as catalysts. The particle size is expected to be <20 μm. Suitability would have to be determined by the researcher.

      Helpful?

  2. this zeolite is a natural zeolite?, i.e., the Clinoptilolite.?. how would I know, if only ZEOLITE is mentioned on the bottle.

    1 answer
    1. This product is of synthetic origin. This information is published in the lot-specific Certificate of Origin. Please see the link below to request a lot-specific Certificate, under the ‘Certificate of Origin’ tab:

      https://www.sigmaaldrich.com/product/sigma/96096#product-documentation

      Helpful?

  3. which type of zeolite it is? Is it natural or synthetic? How would I know if only zeolite is mentioned on it?

    1 answer
    1. This product is of synthetic origin. This information is published in the lot-specific Certificate of Origin. Please see the link below to request a lot-specific Certificate, under the ‘Certificate of Origin’ tab:

      https://www.sigmaaldrich.com/product/sigma/96096#product-documentation

      Helpful?

  4. How many grams of water can this Zeolite hold per gram?

    1 answer
    1. The water absorption capacity of this material is not tested on a lot to lot basis, however it is expected to exceed 23% by weight.  Adsorptive capacities change with temperature

      Helpful?

  5. Wie ist die Temperaturstabilität des Produkts? Welche Maximaltemperatur darf der Zeolith sehen?

    1 answer
    1. The maximum temperature that this product can withstand is not available. The typical temperature range for regeneration is 200-315 degrees Celsius.

      Helpful?

  6. I ordered Zeolite - CAS Number: 1318-02-1 - Product Number: 96096 (Powder, particle size < 20 mm), 500g (euro 196). Could you tell me the specific surface area of this zeolite?

    1 answer
    1. The surface area is estimated to range from 400 - 700 m2/g.

      Helpful?

  7. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  8. What is the powder size distribution (i.e. D10, D50, and D90) of Zeolite 96096?

    1. The particle size distribution of this material is not determined.

      Helpful?

    2. As mentioned in the 'PROPERTIES' section, the particle size is <20 μm.

      Helpful?

  9. Can you tell me the particle size of the zeolite powder (Zeolite - CAS Number: 1318-02-1 - Product Number: 96096)? Thanks

    1 answer
    1. As mentioned in the 'PROPERTIES' section, the particle size is <20 μm

      Helpful?

  10. For Zeolites 96096. I have perform sequential acid-alkaline treatment to enhance the removal percentage of cation heavy metal in water system. Could you provide me the maximum concentration of acid-alkaline treatment, temperature and contact time? Thanks

    1 answer
    1. Unfortunately, there is no protocol available regarding the acid-alkaline treatment for this product. Empirically, it is suggested to use 1 mol/L acid/alkaline solution at room temperature for 30 mins - 2 hrs.

      Helpful?

1–10 of 17 Questions  

Reviews

No rating value

Active Filters