Skip to Content
Merck

Skip To

69780

L-(+)-Lactic acid solution

~40% in H2O

Synonym(s):

(S)-2-Hydroxypropionic acid, Sarcolactic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
UNSPSC Code:
12161700
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1720251
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

concentration

~40% in H2O

impurities

≤1% D-lactic acid

SMILES string

C[C@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1

InChI key

JVTAAEKCZFNVCJ-REOHCLBHSA-N

Other Notes

Aqueous solutions of l(+)-Lactate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
199257L187527714
concentration

~40% in H2O

concentration

in H2O

concentration

30% in H2O (by weight)

concentration

-

Quality Level

100, 200

Quality Level

200

Quality Level

200

Quality Level

200

form

liquid

form

-

form

-

form

-

impurities

≤1% D-lactic acid

impurities

-

impurities

-

impurities

insoluble in ether, complies, methanol, complies (GC), residual solvents, complies, sugar and orther red. Substances, complies, ≤0.0005% heavy metals (as Pb)


Still not finding the right product?

Explore all of our products under L-(+)-Lactic acid solution


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters