Skip to Content
Merck

Skip To

43816

DL-Dithiothreitol solution

BioUltra, Molecular Biology, ~1 M in H2O

Synonym(s):

Cleland′s reagent, (±)-Dithiothreitol, rac-Dithiothreitol, Dithiothreitol, (±)-threo-1,4-Dimercapto-2,3-butanediol solution, DTT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C4H10O2S2
CAS Number:
Molecular Weight:
154.25
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1719757
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

product line

BioUltra

assay

900-1100 mmol/L (Titration molarity)

form

liquid

reaction suitability

reagent type: reductant

concentration

~1 M in H2O

impurities

DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected

pH

2.7-5.0

density

1.04 g/mL at 20 °C

absorption

cut-off at 330 nm

suitability

in accordance for filter test, suitable for molecular biology

storage temp.

2-8°C

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

General description

Dithiothreitol (DTT), a thiol group protectant,[1] is an excellent reagent for preventing the oxidation of SH groups in proteins. DTT solution is a potent and versatile disulfide-reducing agent.[2] It reduces a disulfide bond by forming a six-membrane ring and initiating a thiol-disulfide interchange.

Application

DL-dithiothreitol solution has been used:

  • in the treatment of intestinal explants
  • in homogenate preparation of gill cells for enzymatic assays
  • in protein electrophoresis for western blotting
  • in transglutaminase activity
  • in sample buffer for SDS-PAGE
  • in Western Blot, as a lysis buffer component for single-nucleus RNA-sequencing
  • as a component of protein extraction buffer for cell lysis

Biochem/physiol Actions

DL-dithiothreitol (DTT) is used in a chemical debonding technique, that helps to remove bacteria from biofilms. It can change the structural stability of the GABA(B) (gamma-aminobutyric acid) receptor by disabling the disulfide bonds between four cysteine residues positioned in the GABA(B1(a)) receptor structure.Dithiothreitol (DTT) is broadly utilized in synthetic peptide synthesis and biochemical arrangements of thiol proteins. It can also be used to study protein chemistry topics like enzyme activity and protein folding. DTT explicitly controls the thiol-disulfide exchange response to completely reduce the intra-or between sub-atomic disulfide bonds in biomolecules. Thiols and the cyclic disulfide of DTT are the products of this reaction.

Features and Benefits

  • BioUltra grade - 1M DTT solution, suitable for molecular biology
  • Free from DNases, RNases, phosphatases, and proteases

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
64656343815D9779
grade

Molecular Biology

grade

-

grade

Molecular Biology

grade

Molecular Biology

assay

900-1100 mmol/L (Titration molarity)

assay

-

assay

≥99.5% (RT)

assay

≥98% (HPLC), ≥99% (titration)

product line

BioUltra

product line

-

product line

BioUltra

product line

-

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

impurities

DNases, none detected, phosphatases, none detected, RNases, none detected, proteases, none detected

impurities

-

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected, ≤0.3% 4,5-dihydroxy-1,2-dithiane

impurities

-

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under DL-Dithiothreitol solution


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Suppression of GABA B receptor function in vivo by disulfide reducing agent, dl-dithiothreitol (DTT)
Carai M A M, et al.
Psychopharmacology, 174(2), 283-290 (2004)
Delivery of human immunodeficiency virus vaccine vectors to the intestine induces enhanced mucosal cellular immunity
Wang L, et al.
Journal of Virology, 83(14), 7166-7175 (2009)
In Utero Exposure to Maternal Diabetes Is Associated With Early Abnormal Vascular Structure in Offspring
Dib A, et al.
Frontiers in physiology, 9, 350-350 (2018)



Global Trade Item Number

SKUGTIN
43816-10ML04061832132334
43816-50ML04061838092311
43816-250ML04061832132419

Questions

1–4 of 4 Questions  
  1. After opening DTT ref 43816-50ML, is the product stable? Can it be reused?

    1 answer
    1. Yes, this product is stable until recommended retest date in original container. This product is a prepared solution of 1M DTT in water. In the original sealed container it is quite stable, with an assigned retest schedule of every 2 years. Assuming that the product is handled properly, the material is expected to be stable until at least the retest date reported on the lot specific Certificate of Analysis.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. How stable is this DTT solution ? it is known that DTT is unstable in solution and should be kept under inert atmosphere. How long can I keep it once open ?

    1 answer
    1. This product is a prepared solution of 1M DTT in water. In the original sealed container it is quite stable, with an assigned retest schedule of every 2 years. Assuming that the product is handled properly, the material is expected to be stable until at least the retest date reported on the lot specific Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters