Skip to Content
Merck

Skip To

31397

Dextran from Leuconostoc mesenteroides

Mr ~60,000

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
EC Number:
232-677-5
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Leuconostoc mesenteroides)

Quality Segment

form

powder

mol wt

Mr ~60,000

color

white

mp

483 °C ((901 °F ))

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Application

Dextran is a branched glucan composed of linear α(1→6) linked glucose units and α (1→3) link initiated branches. Dextran ranges in size from 10,000 to 150,000 Kd. Dextrans are used in many applications as volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components.
Use of dextrans as long and hydrophilic spacer arms improves the performance of immobilized proteins acting on macromolecules.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
313983139009184
biological source

(Leuconostoc mesenteroides)

biological source

(leuconostoc mesenteroides)

biological source

(Leuconostoc spp)

biological source

(Leuconostoc spp.)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

color

white

color

white

color

white

color

-

mp

483 °C ((901 °F ))

mp

483  °C ((901 °F ))

mp

483  °C ((901 °F ))

mp

483 °C ((901 °F))

mol wt

Mr ~60,000

mol wt

Mr ~200,000

mol wt

Mr ~70,000

mol wt

Mr ~100,000


Still not finding the right product?


Storage Class

11 - Combustible Solids

ppe

Eyeshields, Gloves, type N95 (US)

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
31397-500G04061833030837
31397-100G04061826684962

Questions

Reviews

No rating value

Active Filters