Skip to Content
Merck

Skip To

16005

Brij® L23

main component: tricosaethylene glycol dodecyl ether

Synonym(s):

Brij 35, C12E23, Polyoxyethylene (23) lauryl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12161900
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

description

non-ionic

form

pellets

CMC

91 μM

mp

39-43 °C

HLB value

16.9

solubility

H2O: 40 mg/mL at 20 °C, clear to slightly hazy, colorless

cation traces

Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤500 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤500 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

functional group

ether, hydroxyl

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C58H118O24/c1-2-3-4-5-6-7-8-9-10-11-13-60-15-17-62-19-21-64-23-25-66-27-29-68-31-33-70-35-37-72-39-41-74-43-45-76-47-49-78-51-53-80-55-57-82-58-56-81-54-52-79-50-48-77-46-44-75-42-40-73-38-36-71-34-32-69-30-28-67-26-24-65-22-20-63-18-16-61-14-12-59/h59H,2-58H2,1H3

InChI key

IEQAICDLOKRSRL-UHFFFAOYSA-N

Application

Brij® L23 is a detergent used for measuring the concentration of carbon black in lung tissues.[1] It has also been used for selective and rapid solubilization of microbial membrane aldehyde dehydrogenase.[2] Additionally, the product has also been used for assessing whether acrolein could lead to pro-enzyme activation by using gelatin zymography.[3]

Biochem/physiol Actions

Brij® L23 also referred to as Brij 35 or Polyoxyethylene (23) lauryl ether is a non-ionic detergent that lowers the non-specific protein adsorption and reduces electroosmotic pumping drastically.[4] This allows the pH to be varied without adversely affecting the flow rate. It also used as a nonionic surfactant that decreases the absorbance of the blank, inhibits turbidity as well as provides linearity.[5]

Other Notes

Non-ionic polyoxyethylene ether.[6] Used for selective and rapid solubilization of microbial membrane aldehyde dehydrogenase.[2]

Legal Information

Brij is a registered trademark of Croda International PLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P1254235989203724
form

pellets

form

solid

form

solid or liquid

form

gel form

mp

39-43 °C

mp

-

mp

48 °C

mp

-

description

non-ionic

description

non-ionic

description

non-ionic

description

Merck USA index - 14, 7558

cation traces

Ca: ≤10 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, K: ≤500 mg/kg, Mn: ≤5 mg/kg, Ni: ≤5 mg/kg, Zn: ≤5 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Na: ≤500 mg/kg, Cr: ≤5 mg/kg, Pb: ≤5 mg/kg, Mg: ≤5 mg/kg

cation traces

-

cation traces

-

cation traces

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

CMC

91 μM

CMC

91 μM

CMC

-

CMC

92 μM


Still not finding the right product?

Explore all of our products under Brij® L23


Storage Class

11 - Combustible Solids

wgk

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Gloves

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



J K Towns et al.
Analytical chemistry, 63(11), 1126-1132 (1991-06-01)
Capillary zone electrophoretic separations of proteins have been achieved by using nonionic surfactant coated capillaries. Capillaries were prepared by derivatization of the silica surface with octadecylsilane followed by the deposition of a layer of nonionic surfactant from an aqueous solution
B T Dumas et al.
Clinica chimica acta; international journal of clinical chemistry, 258(1), 21-30 (1997-02-03)
A rapid and reliable method for measuring serum albumin employing bromcresol green is described. The addition of albumin to a solution of bromcresol green in a 0.075 M succinate buffer pH 4.20 results in an increase in absorbance at 628
Cheol-Hong Lim et al.
Environmental health and toxicology, 28, e2013014-e2013014 (2013-12-05)
This study was conducted to determine whether nano-sized carbon black exposure results in greater damage in high fat diet-induced overweight rats than normal weight ones and to identify the possible causes of any differences. Two groups of Sprague-Dawley rats allocated



Global Trade Item Number

SKUGTIN
16005-250G-F04061838744968
16005-1KG-R04061838744951
16005-1KG-F04061838744944
16005-6X1KG-R04061832489308
16005-25KG-R04061832489292

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters