Skip to Content
Merck

Skip To

36969

trans-1,2-Dichloroethylene

analytical standard

Synonym(s):

trans-1,2-Dichloroethene, trans-Acetylene dichloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
ClCH=CHCl
CAS Number:
Molecular Weight:
96.94
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
205-860-2
Beilstein/REAXYS Number:
1420761
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

vapor pressure

5.16 psi ( 20 °C)

shelf life

limited shelf life, expiry date on the label

expl. lim.

12.8 %

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.446 (lit.)

bp

48 °C (lit.)

mp

−50 °C (lit.)

density

1.257 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cl\C=C\Cl

InChI

1S/C2H2Cl2/c3-1-2-4/h1-2H/b2-1+

InChI key

KFUSEUYYWQURPO-OWOJBTEDSA-N

General description

trans-1,2-dichloroethylene is a chlorinated aliphatic compound, which can undergo bio-degradation by methanotrophic bacteria during simulated aquifer treatment. It can be used as an industrial solvent in the preparations of oils, fats, waxes and also in the manufacture of rubber.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
trans-1,2-Dichloroethylene may be used as an analytical reference standard for the determination of the analyte in water samples by gas chromatography (GC) based analytical techniques.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR3493D6220948597
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

environmental

application(s)

pharmaceutical small molecule

application(s)

-

application(s)

environmental

format

neat

format

-

format

-

format

neat

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

100

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under trans-1,2-Dichloroethylene


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

42.8 °F - closed cup

flash_point_c

6.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Determination of volatile organic hydrocarbons in water samples by solid-phase dynamic extraction
Jochmann MA, et al.
Analytical and Bioanalytical Chemistry, 387(6), 2163-2174 (2007)
On-site and on-line analysis of chlorinated solvents in ground water using pulse introduction membrane extraction gas chromatography (PIME-GC)
San JA, et al.
Journal of Separation Science, 24(7), 599-605 (2001)
Biodegradation of trans-1, 2-dichloroethylene by methane-utilizing bacteria in an aquifer simulator
Moore.TA, et al.
Environmental Science & Technology, 23, 403-406 (1989)



Global Trade Item Number

SKUGTIN
216254-1MG07790788048556
B8806-1MG04061833443286
36969-1G04061831833225

Questions

Reviews

No rating value

Active Filters