Skip to Content
Merck

Skip To

02473

2-Hexanone

analytical standard

Synonym(s):

Butyl methyl ketone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)3COCH3
CAS Number:
Molecular Weight:
100.16
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
209-731-1
Beilstein/REAXYS Number:
1737676
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor pressure

10 mmHg ( 39 °C)

assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.401

bp

127 °C (lit.)

mp

−57 °C (lit.)

density

0.812 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCC(C)=O

InChI

1S/C6H12O/c1-3-4-5-6(2)7/h3-5H2,1-2H3

InChI key

QQZOPKMRPOGIEB-UHFFFAOYSA-N

General description

2-Hexanone (MBK, 2-Oxohexane) is a volatile organic compound, that exists as a clear, colorless liquid with a sharp odor. It dissolves very easily in water, and can evaporate easily into the air as a vapor. It is occasionally found as a volatile component of normal human biofluids. It was used as an industrial hexacarbon solvent, but is no longer used or made owing to its neurotoxic nature leading to various human diseases.

Application

2-Hexanone may be used as an analytical standard for the determination of the analyte in biological samples, starch-based polymers, moldy building materials, and alcoholic matrices by various chromatography techniques.[1][2][3]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR3502W32900247733-U
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

pharmaceutical small molecule

application(s)

flavors and fragrances

application(s)

cleaning products
cosmetics
food and beverages
personal care
petroleum

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

analytical standard

format

neat

format

-

format

-

format

neat

assay

≥99.5% (GC)

assay

-

assay

≥97%

assay

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under 2-Hexanone


signalword

Danger

Hazard Classifications

Flam. Liq. 3 - Repr. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

73.4 °F - closed cup

flash_point_c

23 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Solid-phase extraction and subsequent gas chromatography-mass spectrometry analysis for identification of complex mixtures of degradation products in starch-based polymers
Hakkarainen M, et al.
Journal of Chromatography A, 741(2), 251-263 (1996)
E L White et al.
Biomedical mass spectrometry, 6(4), 169-172 (1979-04-01)
A quantitative method is reported for the determination of n-hexane, 2-hexanone and 2,5-hexanedione in biological tissues by stable isotope dilution using gas chromatography mass spectrometry. n-[2H14]Hexane, 2-[2H5]hexanone and 2,5-[2H10]hexanedione are used as the isotopic diluents. After tissue homogenization and extraction
T.S.S. Dikshith
Handbook of Chemicals and Safety, 19-Apr, 228-228 (2016)



Global Trade Item Number

SKUGTIN
136476-10G04061831818543
02473-5ML04061838617248

Questions

Reviews

No rating value

Active Filters