Skip to Content
Merck

Skip To

P3B030D01

Pellicon® 3 Cassette with Biomax® Membrane

30 kDa, D screen, 0.11 m2

Synonym(s):

Pellicon® 3 Cassette with BIOMAX 30 kDa Membrane, D screen, 0.11 m2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

eCl@ss:
32031623
UNSPSC Code:
23151806
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

material

composite regenerated cellulose (CRC) membrane, polypropylene (Screen (feed)), polypropylene cap (end cap), polypropylene housing

product line

Pellicon®

storage condition

do not freeze

parameter

4-8 L/min-m2 flow rate

technique(s)

tangential flow filtration (TFF): suitable, ultrafiltration: suitable

H

2.3 cm (0.92 in.)

L

20.6 cm (8.1 in.)

filtration area

0.11 m2

size

23 mL , Volume (feed), 7 mL , Volume (permeate)

width

5.6 cm (2.2 in.) , 56 mm

matrix

(Ultracel)

pore size

30 kDa pore size

pH

1-14

storage temp.

2-8°C

General description

Pellicon® 3 cassettes with Biomax membrane are the optimum tangential flow filtration (TFF) devices for the filtration of solutions containing therapeutical proteins, albumin, hormones, vaccines and growth factors. These advanced, high-performance cassettes are ideal for today′s higher titer therapeutic antibodies, higher viscosity feed formulations as well as the more demanding
filtration processes that require higher operating pressures, temperatures and
caustic cleaning regimes.
From small-scale to full-scale production, Pellicon® 3 cassettes are designed for use in research, process scale-up/scale-down, applications development and full-scale manufacturing. The Pellicon® 3 design and automated manufacturing process provides unbeatable performance consistency between cassette sizes. Pellicon® 3 devices also offer greater cassette size selection for improved scale-up and scale-down process development. The streamlined design allows operators to quickly and easily handle, install and remove Pellicon® 3 cassettes. The materials of construction are compatible with a broad range of chemical cleaning.

Application

monoclonal antibodies, recombinant and non-recombinant proteins, vaccines

Features and Benefits

  • Robust, void-free membrane provides optimum product retention and performance consistency
  • Unique feed screen design enables high mass transfer, flux for higher final concentration
  • Fast, reliable scale up/down from lab to production scale
  • Automated manufacturing delivers unbeatable performance consistency and reliability
  • Easy to install and clean
  • Extreme temperature and chemical compatibility

Other Notes

These products are manufactured in a facility which adheres to Good Manufacturing Practices.

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P3C003C01P3B030D00P3B030D05
pore size

30 kDa pore size

pore size

3 kDa pore size

pore size

30 kDa pore size

pore size

30 kDa pore size

technique(s)

tangential flow filtration (TFF): suitable, ultrafiltration: suitable

technique(s)

tangential flow filtration (TFF): suitable, ultrafiltration: suitable

technique(s)

tangential flow filtration (TFF): suitable, ultrafiltration: suitable

technique(s)

tangential flow filtration (TFF): suitable, ultrafiltration: suitable

filtration area

0.11 m2

filtration area

0.11 m2

filtration area

88 cm2

filtration area

0.57 m2

product line

Pellicon®

product line

Pellicon®

product line

Pellicon®

product line

Pellicon®

matrix

(Ultracel)

matrix

(Ultracel)

matrix

(Ultracel)

matrix

(Ultracel)

material

composite regenerated cellulose (CRC) membrane, polypropylene cap (end cap), polypropylene (Screen (feed)), polypropylene housing

material

composite regenerated cellulose (CRC) membrane, polypropylene (Screen (feed)), polypropylene cap (end cap), polypropylene housing

material

composite regenerated cellulose (CRC) membrane, polypropylene (Screen (feed)), polypropylene cap (end cap), polypropylene housing

material

cap (Polypropylene-encapsulated stainless steel end cap), composite regenerated cellulose (CRC) membrane, polypropylene (Screen (feed)), polypropylene housing


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
P3B030D0104055977304596

Questions

Reviews

No rating value

Active Filters