Skip to Content
Merck

Skip To

BM-PACK

Blood typing Bulk Reagent - ABO & RHD (Rh) Specificities

CORE Anti-D Blend (clones TH-28/MS-26) Bulk Reagent plus Diluent Kit

Synonym(s):

Anti-D (clones TH-28/MS-26) Blood Grouping Reagent plus Diluent

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
41116102
Clone:
TH-28/MS-26, monoclonal
Technique(s):
agglutination assay: suitable
Application:
Agglut.
Citations:
-
Note: For further information on price, availability or to place an order, please contact us


biological source

human

antibody form

unpurified

clone

TH-28/MS-26, monoclonal

form

liquid

packaging

kit of

concentration

10-25 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.1-7.3

isotype

IgG, IgM

shipped in

ambient

storage temp.

2-8°C

General description

Reacts with all D weak and D partial red cells, including DVI


Contains 0.1% (w/v) Sodium Azide


Filtered through a 0.22μm terminal filter


Shelf Life: 36 months from date of filtration

Application

For use by Tube;Slide;Microplate;Gel and Column techniques
Reagent + Diluent Kit

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
BM-PACK04053252024320